log in or register
Product Type:
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Process Equipment & Instrumentation > Automation Control > CR Magnetics Split Core Current Transducers
CR Magnetics Split Core Current Transducers
CR Magnetics Split Core Current Transducers
  • A variety of inputs and outputs are available to satisfy any application
  • 35mm DIN Rail or Panel Mount
  • Available with analog 4-20 ma or 0-5 Vdc outputs
  • 24 Vdc powered
  • Use with external current transformers
  • Highest precision available
  • Connection diagram printed on case
  • Directly connect to PLC
  • Sense motor stalls and short circuits
  • Monitor industrial instrumentation
  • Check process control loops
CR Magnetics is proud to introduce a new line of Split Core Current Transducers. Split core devices give the user more options when implementing a sensing application. These devices can be installed on existing power systems without the need to remove power. Also, they make assembling new nstallations much easier than solid core designs. The advanced package includes capability of mounting on either a panel or standard DIN rail installations.
CR4110/20S Series
The CR4110/20S Series provide True RMS sensing of the current waveform. True RMS is recommended when sensing current waveforms that are variable frequency, chopped up, or non-sinusoidal in general.
CR4410/20S Series
The CR4410/20S Series provide Average RMS sensing of the current waveform. Average RMS is recommended when sensing current waveforms that have a fixed frequency, and are usually for loads operated from standard utility power.
CR4210S Series
The CR4210S Series are Self Powered devices providing a proportional DC voltageoutput that is powered by the sensed current. These products are ideal for applications where instrument power is not available.
CR4220S Series
The CR4220S Series are Loop Powered devices providing a proportional DC current output that is powered by a DC power supply within the output current loop. These products are ideal for applications where instrument power is remotely located away from the sensing point.
CR5210/20S Series
The CR5210/20S Series are DC Current Sensing devices that give the average DC value of a direct current input waveform. The CR5210S has a +/- 5VDC that is Bi-Directional, ideal for battery charging applciations.
CR5410S Series
The CR5410S Series are AC/DC Current Sensing devices providing a proportional DC voltage output that provides an identical calibrated waveform to the input current signal. Ideal for closed loop applciations.
 Product Detail
Sort
 Availability
 Price
Representative photo only CR4220S 5 4 to 20 mA DC output AC Avg Loop Powered 0 5 Amps
CR4220S-5, 4 to 20 mA DC output, AC Avg Loop Powered, 0-5 Amps
Product Type Current transducers, split core,Description 4220 Series, single phase, 4 to 20 mA, loop powered,Brand CR Magnetics
EW-68511-14
Usually ships in 8 days.
$143.50 USD/EACH
Qty:
Representative photo only CR5220S 30 4 20 mA DC output DC Current 0 30 Amps
CR5220S-30, 4 - 20 mA DC output, DC Current, 0-30 Amps
Product Type Current transducers, split core,Description 5210 Series DC, 0 to 5 VDC,Brand CR Magnetics
EW-68511-18
Usually ships in 8 days.
$206.50 USD/EACH
Qty:
Representative photo only CR4110S 15 0 5 VDC output AC True RMS 0 15 Amps
CR4110S-15, 0 - 5 VDC output, AC True RMS, 0-15 Amps
Product Type Current transducers, split core,Description 4110 Series, single-phase, 0 to 5 VDC,Brand CR Magnetics
EW-68511-00
Usually ships in 8 days.
$225.00 USD/EACH
Qty:
Representative photo only CR4110S 100 0 5 VDC output AC True RMS 0 100 Amps
CR4110S-100, 0 - 5 VDC output, AC True RMS, 0-100 Amps
Product Type Current transducers, split core,Description 4110 Series, single-phase, 0 to 5 VDC,Brand CR Magnetics
EW-68511-02
Usually ships in 8 days.
$225.00 USD/EACH
Qty:
Representative photo only CR4210S 5 AC Avg Self Powered 0 5 Vdc 0 5 Amps
CR4210S-5, AC Avg., Self Powered, 0-5 Vdc, 0-5 Amps
Product Type Current transducers, split core,Description 4210 Series, single phase, 0 to 5 VDC, self powered,Brand CR Magnetics
EW-68511-04
Usually ships in 13 days.
$121.00 USD/EACH
Qty:
Representative photo only Single Phase 4 20MA Ac Current Average Rms Transducers
Single Phase 4-20MA Ac Current-Average Rms Transducers
Product Type Transducers,Brand CR Magnetics,Model CR4220S-50
EW-68480-04
Call 1-888-358-4705 for availability.
$130.25 USD/EACH
Qty:
Representative photo only CR4110S 50 0 5 VDC output AC True RMS 0 50 Amps
CR4110S-50, 0 - 5 VDC output, AC True RMS. 0-50 Amps
Product Type Current transducers, split core,Description 4110 Series, single-phase, 0 to 5 VDC,Brand CR Magnetics
EW-68511-01
Usually ships in 8 days.
$225.00 USD/EACH
Qty:
Representative photo only CR4120S 10 4 20 mA DC Output True RMS 0 10 Amps
CR4120S-10, 4-20 mA, DC Output, True RMS, 0-10 Amps
Product Type Current transducers, split core,Description 4120 Series, single-phase, 4 to 20 mA,Brand CR Magnetics
EW-68511-03
Usually ships in 8 days.
$234.75 USD/EACH
Qty:
Representative photo only CR4211S 100 0 10 VDC output AC Avg Self Powered 0 100 Amps
CR4211S-100, 0 - 10 VDC output, AC Avg Self Powered, 0-100 Amps
Product Type Current transducers, split core,Description 4211 Series, single phase, 0 to 10 VDC, self powered,Brand CR Magnetics
EW-68511-10
Usually ships in 8 days.
$154.00 USD/EACH
Qty:
Representative photo only CR4120S 5 Split Core 4 20mA 0 5 Amp
CR4120S-5 Split-Core 4-20mA/0-5 Amp
Product Type Current transducers, split core,Description 4120 Series, single-phase, 4 to 20 mA,Brand CR Magnetics
EW-68862-06
Usually ships in 13 days.
$234.75 USD/EACH
Qty:
Representative photo only AC Current Avg RMS Transducer Single Element 0 5VDC Spilt Core
AC Current Avg RMS Transducer Single Element 0-5VDC Spilt Core
Product Type Transducers,Brand CR Magnetics,Model CR4410S-100
EW-68479-25
Usually ships in 7 days.
$148.75 USD/EACH
Qty:
Representative photo only CR4211S 30 0 10 VDC output AC Avg Self Powered 0 30 Amps
CR4211S-30, 0 - 10 VDC output, AC Avg Self Powered, 0-30 Amps
Product Type Current transducers, split core,Description 4211 Series, single phase, 0 to 10 VDC, self powered,Brand CR Magnetics
EW-68511-09
Usually ships in 8 days.
$154.00 USD/EACH
Qty:
Representative photo only CR4210S 30 AC Avg Self Powered 0 5 Vdc 0 30 Amps
CR4210S-30, AC Avg., Self Powered, 0-5 Vdc, 0-30 Amps
Product Type Current transducers, split core,Description 4210 Series, single phase, 0 to 5 VDC, self powered,Brand CR Magnetics
EW-68511-05
Usually ships in 8 days.
$121.00 USD/EACH
Qty:
Representative photo only CR4211S 20 0 10 VDC output AC Avg Self Powered 0 20 Amps
CR4211S-20, 0 - 10 VDC output, AC Avg Self Powered, 0-20 Amps
Product Type Current transducers, split core,Description 4211 Series, single phase, 0 to 10 VDC, self powered,Brand CR Magnetics
EW-68511-08
Usually ships in 8 days.
$154.00 USD/EACH
Qty:
Representative photo only CR5220S 200 4 20 mA DC output DC Current 0 200 Amps
CR5220S-200, 4 - 20 mA DC output, DC Current, 0-200 Amps
Product Type Current transducers, split core,Description 5220 Series DC, 4 to 20 mA,Brand CR Magnetics
EW-68511-19
Usually ships in 8 days.
$205.50 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
Home  >  Process Equipment & Instrumentation > Automation Control > CR Magnetics Split Core Current Transducers
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title