Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Timers (121)
Brand:
Troemner (201)
InnoCal (91)
Product Type:
Availability:
In Stock (148)
Class:
Class 1 (128)
Class 4 (126)
CE Compliance:
Yes (90)
Mass (g):
100 (30)
200 (30)
500 (29)
10 (17)
50 (17)
1000 (11)
150 (10)
300 (10)
400 (10)
2000 (10)
5000 (10)
30 (9)
1 (9)
2 (9)
20 (9)
5 (8)
1500 (8)
3000 (8)
4000 (8)
0.01 (2)
Mass (kg):
10 (15)
20 (14)
25 (9)
30 (6)
5 (6)
2 (5)
1 (4)
50 (3)
3 (2)
100 (2)
4 (1)
35 (1)
Mass (lbs):
2 (8)
50 (7)
25 (7)
1 (6)
5 (5)
10 (5)
20 (5)
0.5 (3)
30 (2)
200 (2)
100 (2)
3 (1)
4 (1)
Applications:
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Calibration

Calibration

Calibration

When you order a new instrument from Cole-Parmer you can also receive calibration services. Simply add the specific calibration service sku to your order to have the item calibrated before it is shipped. It will be ready for use as soon as it arrives at your facility. If you’re instruments need to be recalibrated you can also ship existing instruments to our accredited metrology laboratory for regularly scheduled maintenance calibration services. When your instrument reaches our lab, it is cleaned, calibrated, and quickly returned. If your facility is located in the Chicago area, or if you require a large volume of instrument calibration there is a possibility for an on-site visit. Whatever option you choose, Cole-Parmer offers calibration services for new or existing instruments traceable to NIST-standards.
 Product Detail
Sort
 Availability
 Price
Representative photo only NIST Traceable Calibration 2 4 Point Liquid in Glass Thermometer
EW-17006-03
Usually ships in 8 days.
$160.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration Thermocouple Thermometer System meter probe
EW-17002-10
Usually ships in 7 days.
$160.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration 5 9 Point Liquid in Glass Thermometer
EW-17006-05
Usually ships in 7 days.
$210.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration Pressure Vacuum For gauges with accuracy 0 75 or 0 75
EW-17040-12
Usually ships in 4 days.
$140.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration RTD Meter Probe System

NIST-Traceable Calibration; RTD Meter & Probe System

Product Type Temperature, Meter, Probe, and System Calibrations,Brand InnoCal
EW-17002-04
Usually ships in 7 days.
$160.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration Standard Accuracy Humidity and Temperature
EW-17030-28
Usually ships in 7 days.
$140.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration Hydrometer with test data one point midpoint of Hydrometer s range
EW-17107-00
Usually ships in 7 days.
$130.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration High Accuracy Humidity and Temperature
EW-17030-20
Usually ships in 7 days.
$199.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration RH Temperature Datalogger
EW-17030-24
Usually ships in 9 days.
$175.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration IR Thermometer

NIST-Traceable Calibration; IR Thermometer

Product Type Temperature, Meter, Probe, and System Calibrations,Brand InnoCal
EW-17004-00
Usually ships in 4 days.
$130.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration Datalogger

NIST-Traceable Calibration; Datalogger

Product Type Datalogger
EW-17002-20
Usually ships in 14 days.
$160.00 USD/EACH

Qty:

EW-30008-80 Fluke<small><sup>&reg;</sup></small> 754 Documenting Process Calibrator

Fluke 754 Documenting Process Calibrator

Product Type Process Calibrators/Meters,Brand Fluke,Model FLUKE-754
EW-30008-80
In stock.
$6,299.95 USD/EACH

Qty:

Representative photo only NIST Traceable Report With Data Balances Scales all types
EW-17101-52
Usually ships in 7 days.
$140.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration Handheld Precision Process Calibrator
EW-17101-82
Usually ships in 8 days.
$170.00 USD/EACH

Qty:

Representative photo only NIST Traceable Calibration pH Meter
EW-17106-20
Usually ships in 9 days.
$115.00 USD/EACH

Qty:
See all pH Calibrations

1138 results (showing 1 - 15 )
Per Page 
Home  >  Calibration

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title