Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Home  >  Calibrators
Calibrators
Calibrators
If you have pressure calibrator needs over 10,000 psi here’s something you can try. Use a pressure calibrator with an integral pump and combine it with an external pressure module to calibrate up to 10,000 psi. A pressure calibrator can also be portable. Portable calibrators are used to calibrate pressure gauges, transmitters and transducers. Hand pumps are available as pneumatic or hydraulic pumps. Pneumatic hand pumps generate pressures of up to 600 psi. Hydraulic hand pumps generate pressures up to 10,000 psi with a few squeezes of the pump. These pumps utilize distilled water or hydraulic fluid to achieve high pressures. Deadweight testers are ideal for calibrating, setting, testing and repairing pressure measurement and control devices. Deadweight testers feature two-stage hydraulic pressure generators for pressures up to 10,000 psi. Weights generate the necessary pressure level to test your gauges, transmitters or transducers. Select a pressure calibrator that best fits your applications to keep your pressure gauges accurate and precise. If you need help to choose the right pressure calibrator contact a Cole-Parmer Technical Expert today.
 Product Detail
Sort
 Availability
 Price
GE Druck PM 620 Pressure Module, Absolute/Gauge Pressure; 0 to 15000 psi
  • Product Type: Pressure/Vacuum Calibrators
  • Description: Compatible with noncorrosive gas/fluid and stainless steel
  • Description: Absolute or gauge pressure (unit of measure may be changed on the calibrator)
EW-23001-30
Available for order
or call 1-888-358-4705 to order
$2,331.00 USD/EACH
Qty:
Fluke Calibration P5514 Comparison Test Pump, 0 to 10,000 psi
  • Product Type: Calibration Pumps
  • Description: Hydraulic, bench mount
  • Range: 0 to 10,000 psi (700 bar)
EW-68452-90
Available for order
or call 1-888-358-4705 to order
$1,460.00 USD/EACH
Qty:
Pneumatic pump & vacuum, 600 psi to -28 in Hg
  • Product Type: Calibration Pumps
  • Model: TP1-40
  • Qty/ea: 1
EW-68454-30
Usually ships in 5 days.
$847.00 USD/EACH
Qty:
GE Druck DPI 620 Protective System Carrying Case
  • Product Type: Carrying Case
  • Model: IO620-CASE-2
  • Qty/ea: 1
EW-23001-52
Available for order
or call 1-888-358-4705 to order
$105.00 USD/EACH
Qty:
GE Druck PV212-10K Hydraulic Hand Pump; 10,000 psi
  • Product Type: Calibration Pumps
  • Range: 0 to 10,000 psi
  • Model: PV212
EW-68974-34
Available for order
or call 1-888-358-4705 to order
$961.00 USD/EACH
Qty:
GE Druck DPI 610LP Portable Pressure Calibrator, -5 to 5"WC
  • Product Type: Pressure/Vacuum Calibrators
  • Description: DPI 610 Calibrator
  • Range: +/-5 in WC
EW-68974-50
Available for order
or call 1-888-358-4705 to order
$7,096.00 USD/EACH
Qty:
GE Druck DPI 610LP Portable Pressure Calibrator, -10 to 10"WC
  • Product Type: Pressure/Vacuum Calibrators
  • Description: DPI 610 Calibrator
  • Range: +/-10 in WC
EW-68974-52
Available for order
or call 1-888-358-4705 to order
$7,096.00 USD/EACH
Qty:
GE Druck DPI 615LP Portable Pressure Calibrator, Documenting, +/-10" WC
  • Product Type: Pressure/Vacuum Calibrators
  • Range: +/-10 in WC
  • Media compatibility: liquids or gases compatible with stainless steel
EW-68974-64
Available for order
or call 1-888-358-4705 to order
$7,649.00 USD/EACH
Qty:
GE Druck DPI 610LP Portable Pressure Calibrator, -20 to 20"WC
  • Product Type: Pressure/Vacuum Calibrators
  • Range: +/-20 in WC
  • Media compatibility: liquids or gases compatible with stainless steel
EW-68974-54
Available for order
or call 1-888-358-4705 to order
$7,096.00 USD/EACH
Qty:
Ashcroft Pressure Gauge Comparator, 0-100 psi
  • Product Type: Pressure/Vacuum Calibrators
  • Range: 0-100 psi
  • Operating fluid: SAE 20 weight oil
EW-68840-52
Available for order
or call 1-888-358-4705 to order
$4,953.00 USD/EACH
$4,457.70 USD/EACH
Qty:
GE Druck DPI 615LP Portable Pressure Calibrator, Documenting, +/-20" WC
  • Product Type: Pressure/Vacuum Calibrators
  • Range: +/-20 in WC
  • Media compatibility: liquids or gases compatible with stainless steel
EW-68974-66
Available for order
or call 1-888-358-4705 to order
$7,649.00 USD/EACH
Qty:
GE Druck DPI 610LP Portable Pressure Calibrator, -30 to 30"WC
  • Product Type: Pressure/Vacuum Calibrators
  • Range: +/-30 in WC
  • Media compatibility: liquids or gases compatible with stainless steel
EW-68974-56
Available for order
or call 1-888-358-4705 to order
$7,096.00 USD/EACH
Qty:
Ashcroft Pressure GaugeComparator, 500 spi
  • Product Type: Pressure/Vacuum Calibrators
  • Range: 0-500 psi
  • Operating fluid: SAE 20 wight oil
EW-68840-54
Available for order
or call 1-888-358-4705 to order
$4,953.00 USD/EACH
$4,457.70 USD/EACH
Qty:
GE Druck PM 620 Pressure Module, Gauge Pressure; -14.5 to 30 psi
  • Product Type: Pressure/Vacuum Calibrators
  • Description: Compatible with noncorrosive gas/fluid and stainless steel
  • Description: Gauge pressure (referenced to atmosphere)
EW-23001-06
Available for order
or call 1-888-358-4705 to order
$1,479.00 USD/EACH
Qty:
GE Druck DPI 615LP Portable Pressure Calibrator, Documenting, +/-30" WC
  • Product Type: Pressure/Vacuum Calibrators
  • Range: +/-30 in WC
  • Media compatibility: liquids or gases compatible with stainless steel
EW-68974-68
Available for order
or call 1-888-358-4705 to order
$7,649.00 USD/EACH
Qty:
89 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#
#PRODUCTHEADER#
#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Calibrators