United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Furniture & Fume Hoods
>
Glove Boxes
> Captair Pyramid Glove Bag
Captair Pyramid Glove Bag
Easily set up a controlled environment wherever you need it
Protects you from hazardous materials
Saves lab space
Set up this light-weight pyramid glove bag to provide a controlled environment for user protection and still-air materials handling. Easily transport the lightweight, self-supporting pyramid to any location and assemble in minutes.
Unit features an 18-1/2” (47 cm) side opening for introduction of materials, a zip lock to ensure air-tight sealing, and a port to exhaust/purge gas. Pyramid is constructed from transparent 250
m PVC with frame and 0.2 mm thick butyl gloves. Glove bag is collapsible for easy storage, and can be cleaned with a mild cleanser for continued use. Glove bag cannot be autoclaved.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Captair Pyramid Glove Bag
Product Type Glove Bags,Brand Captair,Model 2200ANM
EW-33666-50
In stock.
$380.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Captair
Pyramid Glove Bag
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Furniture & Fume Hoods
>
Glove Boxes
> Captair Pyramid Glove Bag
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title