This product is now discontinued, View Related Products
CarChip Pro and CarChip Fleet Pro Driving & Engine Performance Data Loggers
Powerful fleet management tool to lower costs and increase productivity
  • Logs time, date, and distance traveled for each trip
  • Speed is logged up to every second
  • Accident logs capture the critical 20 seconds during a hard-braking event
  • CarChip Fleet Pro logger records up to 1200 hours of data before overwriting earliest trip data
The CarChip Fleet Pro plugs directly into the OBD II (onboard diagnostics) port in a car of vehicle where it continuously logs vehicle and driver performance. The CarChip Fleet Pro helps keep fleet vehicles in peak running condition by detecting malfunctions early and taking the guesswork out of diagnosing problems. Log any 4 out of 23 possible engine parameters that affect operation, emissions, and fuel economy. Also has user-settable and audible alarms for speed, hard braking, and quick acceleration.
Perfomance parameters monitored include emissions readiness, RPM, throttle position, engine load, fuel pressure, fuel system status, battery voltage, timing advance, coolant temperature, air flow rate, intake air temperature, intake manifold pressure, short-term fuel trim (2 each), long-term fuel trim (2 each), and oxygen sensor output voltage (8 each).
Requires Fleet Management Software (68511-82 - sold separately) to operate.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Davis Instruments 8246 Engine Performance Logger EW-68511-75 Mfr # 8246
COMPARE
Engine Performance Logger
Call For Price
Davis Instruments 8128 GPS Module (used with 8246 and 8126OBD) EW-68511-76 Mfr # 8128
COMPARE
GPS Module (used with 8246 and 8126OBD)
Call For Price
CarChip 8129 Wireless Download Module (used with 8246 & 8126OBD) EW-68511-77 Mfr # 8129
COMPARE
Wireless Download Module (used with 8246 & 8126OBD)
Call For Price
CarChip 8251 Wirelss Download Interface Module (used with 8246) EW-68511-78 Mfr # 8251
COMPARE
Wirelss Download Interface Module (used with 8246)
Call For Price
CarChip 8130 Wireless Download Base Station (used with 8246 & 8126OBD) EW-68511-80 Mfr # 8130
COMPARE
Wireless Download Base Station (used with 8246 & 8126OBD)
Call For Price
Davis Instruments 8131 Wireless Download Configuration Cable EW-68511-81 Mfr # 8131
COMPARE
Wireless Download Configuration Cable
Call For Price
Davis Instruments 8186 Software For Use With Both 8246 & 8126OBD EW-68511-82 Mfr # 8186
COMPARE
Software For Use With Both 8246 & 8126OBD
Call For Price
Showing 1- 7 of 7