log in or register
Price Range:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Process Equipment & Instrumentation > Steam Handling > Spirax Sarco Thermostatic Steam Traps > Cast iron float and thermostatic steam traps (in-line horizontal configuration)
Cast iron float and thermostatic steam traps (in-line horizontal configuration)
Cast iron float and thermostatic steam traps (in-line horizontal configuration)
  • Drain condensate as quickly as it forms while removing air and other noncondensable gases
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
  • Air heating coils
  • Heat exchangers
  • Steam main drip stations
  • Small process equipment, particularly when controlled by a modulating temperature control valve
  • Replacement of less efficient inverted bucket traps on heating equipment
 Product Detail
Sort
 Availability
 Price
EW-07600-38 Cast iron float and thermostatic steam trap (in-line horizontal configuration); 1/2" NPT(F), 15 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 1/2" NPT(F), 15 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-38
Usually ships in 11 days.
$224.00 USD/EACH
Qty:
Representative photo only Cast iron float and thermostatic steam trap in line horizontal configuration 3 4 NPT F 125 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 3/4" NPT(F), 125 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-52
Usually ships in 7 days.
$363.00 USD/EACH
Qty:
Representative photo only Cast iron float and thermostatic steam trap in line horizontal configuration 3 4 NPT F 15 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 3/4" NPT(F), 15 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-46
Usually ships in 7 days.
$261.00 USD/EACH
Qty:
Representative photo only Cast iron float and thermostatic steam trap in line horizontal configuration 3 4 NPT F 75 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 3/4" NPT(F), 75 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-50
Usually ships in 7 days.
$351.00 USD/EACH
Qty:
Representative photo only Cast iron float and thermostatic steam trap in line horizontal configuration 3 4 NPT F 30 psig
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 3/4" NPT(F), 30 psig
Product Type Steam handling fittings,Material Cast iron,Max temperature 45F (25C) of superheat
EW-07600-48
Usually ships in 7 days.
$261.00 USD/EACH
Qty:
5 results (showing 1 - 5 )
Per Page 
Home  >  Process Equipment & Instrumentation > Steam Handling > Spirax Sarco Thermostatic Steam Traps > Cast iron float and thermostatic steam traps (in-line horizontal configuration)
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title