United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Product Type:
Cell Density Analyzer (Cuvette)
(1)
Cell Density Analyzer (Micro)
(1)
Cell Density Analyzer (Dual)
(1)
[+]
Show more
(1)
Software for Cell Density Analyzer
(1)
[-]
Show less
Price Range:
$100 to $299
(1)
$800 to $1,099
(2)
$1,100 to $1,999
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Life Science
>
Microbiology
> Cell Density Photometers
Cell Density Photometers
Simple and easy to use—dedicated to measuring cell densities in suspension at 600 nm
Analyze samples as small as 20µL volume
No computer required to operate fully built-in controls
Small enough to use inside incubation chambers, anaerobic compartments or other environmentally controlled areas
Use these portable, lightweight cell density meters to measure the growth rates of E. coli, yeast cells and other biological agents in suspension at the 600 nm wavelength of light. Photometers measure % transmission (% T) and logarithmic transmission absorption (Abs), plus feature an intervalometer for time-lapse recording.
The 600 nm LED light focuses on the sample through fiber optic cables to obtain the readings quickly—no warm up time required. A simple touch panel provides eight separate functions and is easily wiped down in the event of spills. The fully recharged battery assembly provides 30 hours of operation; recharge time is 12 hours when the instrument is turned off.
You can easily extract data for further analysis then download results directly to a PC via the built-in USB port. For added convenience, purchase the SpectralaxTM data collection software separately (39459-14) to save data in Excel™ format. This easy-to-use software will automatically prompt you through set up and use. Spectralax is Windows® XP/VISTA/ 7 compatible.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Cuvette sample site cell density photometer, DSM-cuvette, universal power supply
Product Type Cell Density Analyzer (Cuvette),Output RS-232,Display 3-1⁄2-digit LCD
EW-39459-10
Usually ships in 5 days.
$850.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cell Density Photometers
Micro sample site, cell density photometer, universal voltage
Product Type Cell Density Analyzer (Micro),Output RS-232,Display 3-1⁄2-digit LCD
EW-39459-12
Usually ships in 5 days.
$850.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Software for Cell Density Instruments 39459-10,-12,-14
Product Type Software for Cell Density Analyzer,Output RS-232,Display 3-1⁄2-digit LCD
EW-39459-40
Usually ships in 4 days.
$200.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cell Density Photometers
Dual sample site, cell density photometer, universal voltage
Product Type Cell Density Analyzer (Dual) ,Output RS-232,Display 3-1⁄2-digit LCD
EW-39459-14
Usually ships in 5 days.
$1,250.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cell Density Photometers
4
results (showing
1
-
4
)
Per Page
5
10
15
25
50
Home
>
Life Science
>
Microbiology
> Cell Density Photometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title