United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Brand:
Dickson
(44)
Davis
(40)
Partlow
(25)
[+]
Show more
(6)
ABB Commander
(11)
Cole-Parmer
(2)
Honeywell
(2)
Fuji
(1)
Chessell
(1)
Graphic Controls
(1)
[-]
Show less
Product Type:
Chart Recorder Paper
(249)
Chart Recorder Pen
(46)
Print Cartridge
(2)
[+]
Show more
(2)
Chart Recorder Ink
(1)
Hygrothermograph paper
(1)
[-]
Show less
Price Range:
$0 to $100
(297)
$100 to $299
(7)
Availability:
In Stock
(101)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Recorders & Data Acquisition
>
Recorders
> Chart Recorder Paper and Pens
Chart Recorder Paper and Pens
Chart paper and pens to support most of the popular chart recorders
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Thermal Paper for Honeywell DR4500A 12" Digital Recorders
Product Type Chart Recorder Paper,Model 30755317-001,Brand Honeywell
EW-80663-90
In stock.
$55.00 USD/PACKAGE of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Chart Paper for Flatbed Recorders; 100 mm wide, 4 rolls/pk
Product Type Chart Recorder Paper
EW-80550-80
In stock.
$42.00 USD/PACKAGE of 4
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Chart Paper, 14 To 122°F, 7 Day, 2 Hr. Increment 100 Sheets/package
Product Type Chart Recorder Paper
EW-08368-21
In stock.
$26.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Chart Paper, 30-day roll, 6 rolls/bx
Product Type Chart Recorder Paper
EW-08354-95
In stock.
$86.00 USD/BOX of 6
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Paper for Flatbed Recorders; 200 mm wide, 4 rolls/pk
Product Type Chart Recorder Paper
EW-80550-82
In stock.
$45.00 USD/PACKAGE of 4
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Red Pens 60500402: Pack of 5 for Partlow Recorder
Product Type Chart Recorder Pen,Model 60500402,Brand Partlow
EW-49004-98
In stock.
$35.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Purple Pen for Channel 1, Pack of 6
Product Type Chart Recorder Pen,Model 30735489-007
EW-80661-92
In stock.
$48.00 USD/PACKAGE of 6
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Pen, Red
Product Type Chart Recorder Pen,Model 1009600,Brand Dickson
EW-08498-50
In stock.
$14.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Chart paper, -10 to 50°C, 7 day, 2 hr. increment
Product Type Chart Recorder Paper
EW-08368-41
In stock.
$26.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Chart Paper; 200 mm wide, 3 rolls/pk
Product Type Chart Recorder Paper
EW-08386-45
In stock.
$103.00 USD/PACKAGE of 3
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Black Pen
Product Type Chart Recorder Pen,Model F51712A01X01019
EW-80050-71
In stock.
$11.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Paper, 14 To 122°F, 32 Day, 6 Hr. Increment, 25 Sheets/package
Product Type Chart Recorder Paper
EW-08368-23
In stock.
$26.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Chart Paper, 0 To 200, 24 Hr, 60/bx
Product Type Chart Recorder Paper,Min temperature º C -30,Min temperature º F -22
EW-08414-04
In stock.
$36.00 USD/BOX
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Replacement Chart Paper for Honeywell DR4300 10" Process Recorders, 100 sheets/pk
Product Type Chart Recorder Paper,Model 24001661-001,Brand Honeywell
EW-80661-80
In stock.
$28.00 USD/PACKAGE of 100
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
Red Pen for Channel 2, Pack of 6
Product Type Chart Recorder Pen,Model 30735489-002
EW-80661-90
In stock.
$48.00 USD/PACKAGE of 6
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Chart Recorder Paper and Pens
303
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
Recorders & Data Acquisition
>
Recorders
> Chart Recorder Paper and Pens
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title