Home  >  Tubing, and Hose > Hose > Chemfluor Smooth PTFE-Lined Hose Assemblies
Chemfluor Smooth PTFE-Lined Hose Assemblies
Chemfluor Smooth PTFE-Lined Hose Assemblies
Applications: Sanitary or steam transfer, bottle filling, gas analysis, hydraulic lines, extrusion presses, molding and adhesives
Construction
Tube: Chemfluor® PTFE
Reinforcement: 304 stainless steel braid
Advantages: Thicker PTFE tube wall. Superior kink resistance. Excellent vacuum rating. Neutral to taste, color and odor. Nonstick and noncontaminating. Easily cleaned.
Fittings: Permaseal® 316L stainless steel hex male, female JIC, hex male x male JIC union, or female JIC x male adapter
Certifications: FDA-compliant (21 CFR 177.1550), USP Class VI
Temperature range: –100 to 450° F (–73 to 204°C).
 Product Detail
Sort
 Availability
 Price
Representative photo only Smooth PTFE lined hose assembly 3 4 hex male x 3 4 hex male 1 ft length
Smooth PTFE-lined hose assembly, 3/4" hex male x 3/4" hex male, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96552-04
Usually ships in 10 days.
$184.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 4 hex male x 1 4 hex male 1 ft length
Smooth PTFE-lined hose assembly, 1/4" hex male x 1/4" hex male, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96552-01
Usually ships in 10 days.
$43.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 2 JIC female x 1 2 JIC female 10 ft length
Smooth PTFE-lined hose assembly, 1/2" JIC female x 1/2" JIC female, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96553-13
Usually ships in 10 days.
$189.00 USD/EACH
Qty:
EW-96554-03 Smooth PTFE-lined hose assembly, 1/2" JIC female x 1/2" male, 1-ft length
Smooth PTFE-lined hose assembly, 1/2" JIC female x 1/2" male, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96554-03
Usually ships in 10 days.
$139.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 JIC female x 1 JIC female 1 ft length
Smooth PTFE-lined hose assembly, 1" JIC female x 1" JIC female, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96553-05
Usually ships in 10 days.
$248.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 3 4 hex male x 3 4 JIC male 1 ft length
Smooth PTFE-lined hose assembly, 3/4" hex male x 3/4" JIC male, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96555-04
Usually ships in 10 days.
$236.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 2 hex male x 1 2 hex male 10 ft length
Smooth PTFE-lined hose assembly, 1/2" hex male x 1/2" hex male, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96552-13
Usually ships in 15 days.
$189.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 2 hex male x 1 2 JIC male 10 ft length
Smooth PTFE-lined hose assembly, 1/2" hex male x 1/2" JIC male, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96555-13
Usually ships in 10 days.
$223.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 3 8 JIC female x 3 8 male 10 ft length
Smooth PTFE-lined hose assembly, 3/8" JIC female x 3/8" male, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96554-12
Usually ships in 10 days.
$184.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 4 hex male x 1 4 JIC male 10 ft length
Smooth PTFE-lined hose assembly, 1/4" hex male x 1/4" JIC male, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96555-11
Usually ships in 10 days.
$114.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 3 8 JIC female x 3 8 JIC female 10 ft length
Smooth PTFE-lined hose assembly, 3/8" JIC female x 3/8" JIC female, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96553-12
Usually ships in 11 days.
$151.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 4 hex male x 1 4 JIC male 1 ft length
Smooth PTFE-lined hose assembly, 1/4" hex male x 1/4" JIC male, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96555-01
Usually ships in 10 days.
$56.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 4 hex male x 1 4 hex male 10 ft length
Smooth PTFE-lined hose assembly, 1/4" hex male x 1/4" hex male, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96552-11
Usually ships in 12 days.
$100.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 3 8 hex male x 3 8 JIC male 1 ft length
Smooth PTFE-lined hose assembly, 3/8" hex male x 3/8" JIC male, 1-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96555-02
Usually ships in 10 days.
$74.00 USD/EACH
Qty:
Representative photo only Smooth PTFE lined hose assembly 1 JIC female x 1 male 10 ft length
Smooth PTFE-lined hose assembly, 1" JIC female x 1" male, 10-ft length
Product Type Hose fittings and accessories,Maximum temperature º F 450,Minimum temperature º F -100
EW-96554-15
Usually ships in 10 days.
$602.00 USD/EACH
Qty:
40 results (showing 1 - 15 )
Per Page 
Home  >  Tubing, and Hose > Hose > Chemfluor Smooth PTFE-Lined Hose Assemblies
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title