Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Dichlor (30)
Brand:
Product Type:
Price Range:
Class:
Solids (70)
Purity Grade:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 

Chemicals D

Chemicals D
Choose from our comprehensive line of Chemicals Da to Dy and other Chemicals products.
 Product Detail
Sort
 Availability
 Price

N,N-Diethylhydroxylamine 97% (100g)

  • Product Type: Organic Chemicals
  • CAS Number: 3710-84-7
  • Form: Solid
EW-88107-38
Usually ships in 2 days.
$41.00 USD/EACH

Qty:

Dimethyl Sulfoxide (4 liter) CAS 67-68-5

  • Product Type: Organic Chemicals
  • CAS Number: 67685
  • Form: Liquid
EW-88007-87
Usually ships in 6 days.
$385.00 USD/EACH
$324.99 USD/EACH

Qty:

Dichlorofluorescein TS, 0.1% (w/v) in Dilute Alcohol (120 mL)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS or USP or NF grade
  • CAS Number: Includes 76-54-0, 64-17-5, 67-56-1, 1310-73-2 and 7732-18-5
EW-86990-29
Available for order
or call 1-888-358-4705 to order
$44.75 USD/EACH

Qty:

Dowex 50WX8-100 ion-exchange resin (100g)

  • Product Type: Organic Chemicals
  • CAS Number: 11119-67-8
  • Form: Resin
EW-88305-08
Usually ships in 4 days.
$84.75 USD/EACH

Qty:

Dimethyl Sulfoxide, ICN (100 mL) CAS 67-68-5

  • Product Type: Organic Chemicals
  • CAS Number: 67685
  • Form: Liquid
EW-88058-50
Usually ships in 3 days.
$47.50 USD/EACH
$39.99 USD/EACH

Qty:

Diisopropylamine 99% (2.5l) CAS 108-18-9

  • Product Type: Organic Chemicals
  • CAS Number: 108-18-9
  • Form: Liquid
EW-88150-03
Usually ships in 2 days.
$129.00 USD/EACH

Qty:

2,6-Dichloroindophenol Sodium Salt (Certified ACS) (5g)

  • Product Type: Organic Chemicals
  • Purity grade: ACS Reagent Grade
  • CAS Number: 620451
EW-88080-63
Usually ships in 3 days.
$244.00 USD/EACH

Qty:

Dimethyl Sulfoxide (1 liter) CAS 67-68-5

  • Product Type: Organic Chemicals
  • CAS Number: 67685
  • Form: Liquid
EW-88007-85
Usually ships in 4 days.
$135.00 USD/EACH

Qty:

Drierite, 10-20 mesh, with indicator (500g)

  • Product Type: Inorganic Chemicals
  • CAS Number: 7778-18-9
  • Formula weight: 136.13
EW-88320-83
Usually ships in 9 days.
$64.50 USD/EACH

Qty:

DPD Indicator Solution, for Residual Chlorine Analysis (500 mL)

  • Product Type: Inorganic Chemicals
  • Purity grade: Components all ACS or Reagent grade
  • CAS Number: Includes 6381-92-6, 6283-63-2, 7664-93-9 and 7732-18-5
EW-86971-90
Usually ships in 7 days.
$59.00 USD/EACH

Qty:

N,N-Dimethylformamide (Synthesis) (4 liter)

  • Product Type: Organic Chemicals
  • Purity grade: Synthesis
  • CAS Number: 68-12-2
EW-88056-48
Usually ships in 2 days.
$296.00 USD/EACH

Qty:

4,5-Dihydroxynaphthalene-2,7-disulfonic acid, reagent ACS (10g)

  • Product Type: Organic Chemicals
  • CAS Number: 5808-22-0
  • Form: Powder
EW-88276-68
Usually ships in 5 days.
$67.25 USD/EACH

Qty:

Dicyandiamide 99.5% (2.5kg) CAS 461-58-5

  • Product Type: Organic Chemicals
  • CAS Number: 461585
  • Form: Solid
EW-88106-35
Usually ships in 2 days.
$70.50 USD/EACH

Qty:

N,N'-Dicyclohexylcarbodiimide 99% (100g)

  • Product Type: Organic Chemicals
  • CAS Number: 538-75-0
  • Form: Solid
EW-88106-48
Usually ships in 2 days.
$70.00 USD/EACH
$53.99 USD/EACH

Qty:

Diethanolamine 99% (1kg) CAS 111-42-2 (2,2'-Iminodiethanol)

  • Product Type: Organic Chemicals
  • CAS Number: 111422
  • Form: Solid
EW-88106-52
Usually ships in 2 days.
$66.00 USD/EACH

Qty:

211 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#