Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Chemicals (7882)
Brand:
Product Type:
Availability:
In Stock (48)
Shop Items By:
Class:
Solids (846)
Normality:
Purity Grade:
Certified (292)
ACS (220)
Volume (Liters):
0 to 3 (14)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  > 

Chemicals, Reagents & Lubricants

Chemicals, Reagents & Lubricants
It goes without saying you should be cautious when buying chemicals online. But what should you look out for the most? First you want to make sure that the group offering chemicals for sale is a reputable company. Second, you want to make sure you’re getting the right chemicals. Cole-Parmer has a Material Safety Data Sheet that gives buyers the information on chemicals sold and their hazards and uses. Finally if you want blended substances it’s best to have an expert help you with that such as a Cole-Parmer Technical Expert. Also, reference our Chemical Compatibility Database to evaluate the compatibility of your container or materials with a particular chemical.
Need
Assistance?

Get help from
our Experts!
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price

Cole-Parmer Micro-90 cleaning solution, 1 liter bottle

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Description: One 1 liter bottle
EW-18100-05
Usually ships in 4 days or sooner.
$13.00 USD/EACH

Qty:

Cole-Parmer Micro-90 cleaning solution, case of twelve 1 liter bottles

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Description: Case of twelve 1-liter bottles
EW-18100-04
In stock.
$140.00 USD/CASE

Qty:

Alconox 1104-1 cleaner for manual or ultrasonic cleaning, 4 lb box

  • Product Type: Equipment Cleaner
  • Format: Powder
  • Packaging: 4 lb box
EW-17775-05
In stock.
$40.75 USD/EACH

Qty:

Cole-Parmer Micro-90 cleaning solution, case of four 4 Liter bottles

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Description: Case of four 4-Liter Bottles
EW-18100-11
In stock.
$158.00 USD/CASE

Qty:

Hardman 4001-BG10 Fast-Setting Epoxy Pouches, Amber, 3.5g; 10/Pack

  • Product Type: Adhesives & Epoxies
  • Temperature range (º F): -20 to 180°F
  • Curing time: 24 hours
EW-08778-02
In stock.
$22.25 USD/PACKAGE

Qty:

Cole-Parmer Micro-90 cleaning solution, 5 gallon bottle

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Description: Case of one 5-gal. Bottles
EW-18100-20
In stock.
$292.00 USD/EACH

Qty:

Liqui-Nox Cleaning Detergent, one gallon bottle

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Volume: 1 gal.
EW-17778-40
In stock.
$87.25 USD/EACH

Qty:

Liberty Deionized Water Diluent; 1 gallon

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Volume: 1 gal.
EW-09411-79
Usually ships in 8 days or sooner.
$20.25 USD/EACH
$17.21 USD/EACH

Qty:

ORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (500 mL)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • CAS Number: Includes 7447-40-7, 13746-66-2, 14459-95-1 and 7732-18-5
EW-86974-09
Available for order
or call 1-888-358-4705 to order
$35.25 USD/EACH

Qty:

Liqui-Nox Cleaning Detergent, case of four 1-gallon bottles

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Volume: 1 gal.
EW-17775-40
In stock.
$273.00 USD/CASE of 4

Qty:

Liqui-Nox cleaner for general purpose cleaning, 1 quart bottle

  • Product Type: Equipment Cleaner
  • Format: Liquid
  • Volume: 0.95 Liters
EW-17778-05
In stock.
$29.50 USD/EACH

Qty:

Cole-Parmer Isopropyl Alcohol (IPA), ACS Grade, 4 L

  • Product Type: Organic Chemicals
  • Purity grade: ACS
  • CAS Number: 67-63-0
EW-88405-69
Usually ships in 7 days.
$103.00 USD/EACH

Qty:

Sodium Hydroxide, 0.100 Normal (N/10) (1 L)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • Normality: 0.100
EW-88400-83
Usually ships in 7 days.
$25.00 USD/EACH

Qty:

Sodium Hydroxide, 1.00 Normal (1 gal) (Liquid) 88400-88

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • Normality: 1.00
EW-88400-88
Usually ships in 7 days.
$52.00 USD/EACH

Qty:

Sodium Hydroxide, 0.100 Normal (N/10) (1 gal)

  • Product Type: Inorganic Chemical
  • Purity grade: Components all ACS grade
  • Normality: 0.100
EW-88400-82
Usually ships in 7 days.
$52.25 USD/EACH

Qty:

8192 results (showing 1 - 15 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >