Home  >  Clean Room > Cleanroom Furniture
Cleanroom Furniture
Cleanroom Furniture
What's the best quality to look for in a cleanroom chair? Like all cleanroom furniture, a cleanroom chair is ideal for use in semiconductor, bioscience, pharmaceutical, and board manufacturing. Pair it with other cleanroom products, and you've got a surefire way to keep your particle count down.
Any cleanroom chair should comply with ISO cleanliness standards and each one we carry has been been tested by an independent testing laboratory to assure compliance. Cole-Parmer has cleanroom chairs with vinyl or polyurethane seat materials and base materials of aluminum or plated tubular steel. Added are pneumatic height adjustment and 360° swivel for comfortable positioning. We also have cleanroom stools with vinyl seat materials and pneumatic height adjustment. Both chairs and stools are available in ESD-safe versions, including drag chains and ESD mushroom glides instead of casters. Couple your cleanroom chair with a cleanroom table constructed of 16-gauge type 304 stainless steel available as a flat table and versions with rear splash guard.
 Product Detail
Sort
 Availability
 Price
EW-86309-32 Clean Room Polyurethane Chair, 22" - 32" Height
Clean Room Polyurethane Chair, 22" - 32" Height
Product Type Cleanroom Chair,Material Polyurethane,Brand Bevco
EW-86309-32
In stock.
$340.00 USD/EACH
Qty:
Representative photo only Stainlesss Steel Worktables with Rear Splash 96 L x 30 W
Stainlesss Steel Worktables with Rear Splash, 96"L x 30"W
Product Type Worktables,Model KMS-308
EW-47620-86
Usually ships in 13 days.
$2,087.00 USD/EACH
Qty:
Representative photo only Cleanroom Chair Deluxe Vinyl 17 to 22 25 height ISO Class 7 aluminum base
Cleanroom Chair, Deluxe Vinyl, 17" to 22.25" height, ISO Class 7, aluminum base
Product Type Deluxe Vinyl Cleanroom Chair,Brand Bevco,Model 9050M-C4 BLK
EW-86309-50
Call 1-888-358-4705 for availability.
$285.00 USD/EACH
Qty:
EW-86309-05 Clean Room Vinyl Chair, 23" - 33" Height
Clean Room Vinyl Chair, 23" - 33" Height
Product Type Vinyl Cleanroom Chair,Material Vinyl,Brand Bevco
EW-86309-05
Call 1-888-358-4705 for availability.
$425.00 USD/EACH
Qty:
Representative photo only Cleanroom Chair Deluxe Vinyl 20 to 27 5 height ISO Class 7 aluminum base
Cleanroom Chair, Deluxe Vinyl, 20" to 27.5" height, ISO Class 7, aluminum base
Product Type Deluxe Vinyl Cleanroom Chair,Brand Bevco,Model 9350M-C4 BLK
EW-86309-53
Call 1-888-358-4705 for availability.
$340.00 USD/EACH
Qty:
Representative photo only Stainless Steel work table 36 L x 24 W
Stainless Steel work table, 36"L x 24"W
Product Type Worktables,Model U3SLC-24
EW-86519-00
Call 1-888-358-4705 for availability.
$1,199.00 USD/EACH
Qty:
Representative photo only Clean Room Polyurethane Chair 16 21 Height
Clean Room Polyurethane Chair, 16" - 21" Height
Product Type Cleanroom Chair,Material Polyurethane,Brand Bevco
EW-86309-30
Call 1-888-358-4705 for availability.
$310.00 USD/EACH
Qty:
EW-86520-05 Cleanroom stool, vinyl, 24
Cleanroom stool, vinyl, 24" to 34", aluminum base
Product Type Cleanroom Stool,Material Vinyl,Brand Bevco
EW-86520-05
Call 1-888-358-4705 for availability.
$270.00 USD/EACH
Qty:
Representative photo only Cleanroom Chair ESD Polyurethane 22 to 32 height aluminum base
Cleanroom Chair, ESD Polyurethane, 22" to 32" height, aluminum base
Product Type ESD Cleanroom Chair,Brand Bevco
EW-86309-35
Call 1-888-358-4705 for availability.
$434.00 USD/EACH
Qty:
Representative photo only ESD Clean Room Vinyl Chair 23 33 Height
ESD Clean Room Vinyl Chair, 23" - 33" Height
Product Type ESD Vinyl Cleanroom Chair,Material Vinyl,Brand Bevco
EW-86309-15
Call 1-888-358-4705 for availability.
$446.00 USD/EACH
Qty:
Representative photo only Stainless Steel Worktables with Rear Splash 72 L x 30 W
Stainless Steel Worktables with Rear Splash, 72"L x 30"W
Product Type Worktables,Model KMS-306
EW-47620-84
Usually ships in 9 days.
$1,538.00 USD/EACH
Qty:
Representative photo only Cleanroom Chair Deluxe Vinyl 20 to 27 5 height ISO Class 4 aluminum base
Cleanroom Chair, Deluxe Vinyl, 20" to 27.5" height, ISO Class 4, aluminum base
Product Type Deluxe Vinyl Cleanroom Chair,Brand Bevco,Model 9350M-C1 BLK
EW-86309-55
Call 1-888-358-4705 for availability.
$420.00 USD/EACH
Qty:
Representative photo only Stainless Steel Worktables with Rear Splash 60 L x 30 W
Stainless Steel Worktables with Rear Splash, 60"L x 30"W
Product Type Worktables,Model KMS-305
EW-47620-82
Usually ships in 7 days.
$1,429.00 USD/EACH
Qty:
Representative photo only Cleanroom stool vinyl 25 to 30 chrome tubular steel base
Cleanroom stool, vinyl, 25" to 30", chrome tubular steel base
Product Type Cleanroom Stool,Material Vinyl,Brand Bevco
EW-86520-02
Call 1-888-358-4705 for availability.
$235.00 USD/EACH
Qty:
Representative photo only Cleanroom Stool Vinyl 20 to 27 height aluminum base
Cleanroom Stool, Vinyl, 20" to 27" height, aluminum base
Product Type Cleanroom Stool,Brand Bevco
EW-86520-03
Call 1-888-358-4705 for availability.
$270.00 USD/EACH
Qty:
50 results (showing 1 - 15 )
Per Page 
Home  >  Clean Room > Cleanroom Furniture
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title