Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Price Range:
Availability:
Gel size (cm L):
8 (9)
Gel size (cm W):
8.5 (9)
No. of wells:
12 (7)
17 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Life Science > Electrophoresis Products > Reagents > ClearPAGE Electrophoresis Precast Gels

ClearPAGE Electrophoresis Precast Gels

ClearPAGE Electrophoresis Precast Gels

  • Available for sale only in the United States
  • Quick and easy-to-use precast gels eliminate the need for traditional handcasting
  • Compatible with combination vertical systems (see "Related Product" ad at bottom)
  • Gel technology features a unique polymer matrix for exceptional protein seperation
  • Simple and reuseable — saving time and money
  • Long shelf life saves your lab money
No need to throw away expired gels! These convenient precast gels have over a 12-month shelf life. No change in separation when stored properly during that period. Incredibly strong gels provide high resolution results and are highly resistant to tearing and cracking, even during the drying process. One-of-a-kind polyacrylamide gel with agarose formulation also reduces shrinkage and swelling, resulting in better resolution and more efficient transfer. Use precast gels with buffers sold under "Accessories"—they are not compatible with standard lab buffers. Large sample volumes will still product sharp bands. Cassette design allows longer pathlength, increasing resolution. All gels are 1-mm thick and use a red loading area for easy visualization of wells. Available in fixed or gradient acrylamide concentrations.
 Product Detail
Sort
 Availability
 Price
Representative photo only Instant Blue Stain 1 liter

Instant Blue Stain, 1 liter

Product Type Electrophoresis Accessories
EW-28570-59
In stock.
$209.00 USD/EACH

Qty:

Representative photo only LDS Sample Buffer 10ml 4x

LDS Sample Buffer 10ml, 4x

Product Type Electrophoresis Accessories
EW-28570-49
Usually ships in 6 days.
$19.00 USD/EACH

Qty:

Representative photo only ClearPAGE SDS Gel 4 20 Acrylamide 12 well 10 per Package

ClearPAGE SDS Gel, 4-20% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-44
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only ClearPAGE SDS Gel 16 Acrylamide 12 well 10 per Package

ClearPAGE SDS Gel, 16% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-38
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only Turbo Buffer 50mL

Turbo Buffer 50mL

Product Type Electrophoresis Accessories
EW-28570-53
Usually ships in 6 days.
$11.00 USD/EACH

Qty:

Representative photo only ClearPAGE SDS Gel 12 Acrylamide 17 well 10 per Package

ClearPAGE SDS Gel, 12% Acrylamide, 17 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 17,Gel size cm W 8.5
EW-28570-36
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only Turbo Buffer 500 mL

Turbo Buffer 500 mL

Product Type Electrophoresis Accessories
EW-28570-55
Usually ships in 6 days.
$66.00 USD/EACH

Qty:

Representative photo only ClearPAGE SDS Gel 10 Acrylamide 12 well 10 per Package

ClearPAGE SDS Gel, 10% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-32
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only SDS Non Reducing Run Buffer 50ml 20x

SDS-Non Reducing Run Buffer 50ml, 20x

Product Type Electrophoresis Accessories
EW-28570-51
Usually ships in 6 days.
$11.00 USD/EACH

Qty:

Representative photo only ClearPAGE SDS Gel 4 20 Acrylamide 17 well 10 per Package

ClearPAGE SDS Gel, 4-20% Acrylamide, 17 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 17,Gel size cm W 8.5
EW-28570-46
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

EW-28570-30 ClearPAGE<small><sup>&#153;</sup></small> SDS Gel, 8% Acrylamide, 12 well, 10 per Package

ClearPAGE SDS Gel, 8% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-30
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only ClearPAGE SDS Gel 12 Acrylamide 12 well 10 per Package

ClearPAGE SDS Gel, 12% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-34
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only ClearPAGE SDS Gel 4 12 Acrylamide 12 well 10 per Package

ClearPAGE SDS Gel, 4-12% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-42
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

Representative photo only TGS Transfer Buffer 500ml 10x

TGS Transfer Buffer 500ml, 10x

Product Type Electrophoresis Accessories
EW-28570-57
Usually ships in 6 days.
$22.00 USD/EACH

Qty:

Representative photo only ClearPAGE SDS Gel 4 8 Acrylamide 12 well 10 per Package

ClearPAGE SDS Gel, 4-8% Acrylamide, 12 well, 10 per Package

Product Type Electrophoresis Accessories,No. of wells 12,Gel size cm W 8.5
EW-28570-40
Usually ships in 6 days.
$96.00 USD/PACKAGE

Qty:

15 results (showing 1 - 15 )
Per Page 
Home  >  Life Science > Electrophoresis Products > Reagents > ClearPAGE Electrophoresis Precast Gels

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title