log in or register
Price Range:
Feet per pack:
Sterilizable:
Yes (19)
Tubing OD (mm):
12.8 (3)
11.2 (3)
6.4 (2)
3.2 (1)
4.8 (1)
19.1 (1)
4 (1)
8 (1)
2.4 (1)
5.6 (1)
2.1 (1)
28.6 (1)
9.6 (1)
17.6 (1)
16.0 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Tubing, and Hose > Tubing > Flexible Tubing > Clear C-FLEX Tubing
Clear C-FLEX Tubing
Clear C-FLEX Tubing
Applications: Pharmaceutical, biotech, and general-purpose lab uses requiring high performance. Economical alternative to platinum silicone.
Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent.
Certification: Product is manufactured in accordance with quality regulations CFR Title 21, Part 820 Quality System Requirements (GMP).
Temperature range: -40 to 104°F (–40 to 40°C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
 Product Detail
Sort
 Availability
 Price
Representative photo only Clear C FLEX Tubing 1 2 ID x 3 4 OD 25 ft pack
Clear C-FLEX Tubing, 1/2"ID x 3/4"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-18
In stock.
$137.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 3 8 ID x 5 8 OD 25 ft pack
Clear C-FLEX Tubing, 3/8"ID x 5/8"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-16
In stock.
$97.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 3 4 x 1 1 8 15 ft pk
Clear C-FLEX Tubing, 3/4 x 1-1/8", 15 ft/pk
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-21
In stock.
$269.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 8 ID x 1 4 OD 25 ft pack
Clear C-FLEX Tubing, 1/8"ID x 1/4"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-05
In stock.
$32.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 4 ID x 7 16 OD 25 ft pack
Clear C-FLEX Tubing, 1/4"ID x 7/16"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-11
In stock.
$63.00 USD/PACKAGE
Qty:
EW-06422-10 Clear C-FLEX<small><sup>&reg;</sup></small> tubing, 1/4
Clear C-FLEX Tubing, 1/4"ID x 3/8"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-10
In stock.
$42.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 3 16 ID x 5 16 OD 25 ft pack
Clear C-FLEX Tubing, 3/16"ID x 5/16"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-08
In stock.
$38.80 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 16 ID x 1 8 OD 25 ft pack
Clear C-FLEX Tubing, 1/16"ID x 1/8"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-02
In stock.
$19.50 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 3 32 ID x 5 32 OD 25 ft pack
Clear C-FLEX Tubing, 3/32"ID x 5/32"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-03
In stock.
$26.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 50 ID x 1 12 OD 25 ft pack
Clear C-FLEX Tubing, 1/50"ID x 1/12"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-00
In stock.
$23.50 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 8 ID x 3 16 OD 25 ft pack
Clear C-FLEX Tubing, 1/8"ID x 3/16"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-04
In stock.
$27.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 3 8 ID x 1 2 OD 25 ft pack
Clear C-FLEX Tubing, 3/8"ID x 1/2"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-15
In stock.
$57.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 5 16 ID x 1 2 OD 25 ft pack
Clear C-FLEX Tubing, 5/16"ID x 1/2"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-14
In stock.
$68.00 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 32 ID x 3 32 OD 25 ft pack
Clear C-FLEX Tubing, 1/32"ID x 3/32"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-01
In stock.
$18.50 USD/PACKAGE
Qty:
Representative photo only Clear C FLEX Tubing 1 4 ID x 1 2 OD 25 ft pack
Clear C-FLEX Tubing, 1/4"ID x 1/2"OD, 25 ft/pack
Product Type Tubing,Maximum temperature º F 104,Minimum temperature º F -40
EW-06422-12
In stock.
$75.00 USD/PACKAGE
Qty:
20 results (showing 1 - 15 )
Per Page 
Home  >  Tubing, and Hose > Tubing > Flexible Tubing > Clear C-FLEX Tubing
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title