log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Flow, Level & Valves > Flowmeters > Variable Area Flowmeters > Cole-Parmer Direct Reading Panel-Mount Flowmeters > Cole-Parmer 150-mm Direct Reading Panel-Mount Flowmeters
Cole-Parmer 150-mm Direct Reading Panel-Mount Flowmeters
Cole-Parmer 150-mm Direct Reading Panel-Mount Flowmeters
  • Front shields magnify scale 16%—easy to read from a distance
These direct reading flowmeters have a ceramic millimeter scale is fused onto the flowtube. A vertical tangential locator line ensures hairline accuracy in reading float position. The front polycarbonate shields are magnified 16% for easy, accurate readings.
Select a flowmeter that best suits your application from the several valve options and wetted materials. Nonwetted parts for all models include aluminum frame side plates, polycarbonate shields, and acrylic back plates.
Units are supplied with either a black glass (G), a 316 stainless steel (SS), a sapphire (Sa), or a carboloy ©float (see tables). Direct reading flowmeters for argon, CO2, helium, nitrogen, and oxygen are available—call our Applications Specialists for more information.
Inlet and outlet connections are 1/8" NPT(F) with external threading for panel mounting (two retaining nuts are included).
Flowmeter Options
Aluminum is economical and good for general use with noncorrosive gases and liquids. Aluminum fittings, Buna N O-rings, borosilicate glass flowtubes, 316 stainless steel (SS) 150-mesh inlet screens, and glass or 316 SS floats.
Brass is economical and good for use with water. Chrome-plated brass fittings, Buna N O-rings, borosilicate glass flowtubes, 316 SS 150-mesh inlet screens, and glass or 316 SS floats.
316 Stainless Steel withstands higher temperatures and pressures; and features excellent chemical compatibility. Fittings are 316 SS. Viton® O-rings, borosilicate glass flowtubes, 316 SS 150-mesh inlet screens, and glass or 316 SS floats.
Valve Options include valveless or with valve. Valves come mounted at the inlet (bottom) of applicable flowmeters but can be reconfigured to mount at the outlet. For positive pressure applications with liquids and gases, position the valve at the inlet; for vacuum applications, position at the outlet.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Aluminum Carboloy Float For Air With Valve 100 mL min
Cole-Parmer 150-mm Direct Reading Flowmeter,Aluminum/Carboloy Float; For Air, With Valve; 100 mL/min
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32003-04
In stock.
$174.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Aluminum Ss Float For Air With Valve 200 mL min
Cole-Parmer 150-mm Direct Reading Flowmeter, Aluminum/Ss Float; For Air, With Valve; 200 mL/min
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32003-06
In stock.
$174.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Ss Glass Float For Water With Valve 100 mL min
Cole-Parmer 150-mm Direct Reading Flowmeter, Ss/Glass Float; For Water, With Valve; 100 mL/min
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32005-34
In stock.
$289.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Ss Ss Float For Water With Valve 500 mL min
Cole-Parmer 150-mm Direct Reading Flowmeter, Ss/Ss Float; For Water, With Valve; 500 mL/min
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32005-38
In stock.
$289.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter For Water 316 Ss With Valve 1 2 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter For Water; 316 Ss With Valve, 1.2 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32035-40
In stock.
$299.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter For Air 316 Ss With Valve 1 25 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter For Air; 316 Ss With Valve, 1.25 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32035-10
Usually ships in 14 days.
$289.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Ss Glass Float For Air With Valve 10 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter, Ss/Glass Float; For Air, With Valve; 10 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32005-14
Usually ships in 11 days.
$289.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Aluminum Glass Float For Air With Valve 2 5 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, With Valve; 2.5 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32003-10
In stock.
$174.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter For Water 316 Ss Without Valve 1 2 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter For Water; 316 Ss Without Valve, 1.2 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32032-40
Usually ships in 11 days.
$270.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Aluminum Glass Float For Air With Valve 8 25 scfh
Cole-Parmer 150-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, With Valve; 8.25 scfh
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32003-18
Usually ships in 11 days.
$174.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Ss Carboloy Float For Air Without Valve 100 mL min
Cole-Parmer 150-mm Direct Reading Flowmeter,Ss/Carboloy Float; For Air, Without Valve; 100 mL/min
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32002-04
Usually ships in 11 days.
$270.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter For Air Brass With Valve 23 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter For Air; Brass With Valve, 23 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32034-14
Usually ships in 11 days.
$207.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Ss Sapphire Float For Air With Valve 5 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter, Ss/Sapphire Float; For Air, With Valve; 5 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32005-12
Usually ships in 11 days.
$289.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Brass Sapphire Float For Air With Valve 5 LPM
Cole-Parmer 150-mm Direct Reading Flowmeter, Brass/Sapphire Float; For Air, With Valve; 5 LPM
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32004-12
Usually ships in 11 days.
$195.00 USD/EACH
Qty:
Representative photo only Cole Parmer 150 mm Direct Reading Flowmeter Air Aluminum 316Ss Float For Air 800 mL min
Cole-Parmer 150-mm Direct Reading Flowmeter,Air,Aluminum/316Ss Float; For Air, 800 mL/min
Product Type Variable Area Flowmeter,Description Variable Area, Direct Reading Flowmeter,Max operating temperature º F 250
EW-32030-09
Usually ships in 11 days.
$159.00 USD/EACH
Qty:
148 results (showing 1 - 15 )
Per Page 
Home  >  Flow, Level & Valves > Flowmeters > Variable Area Flowmeters > Cole-Parmer Direct Reading Panel-Mount Flowmeters > Cole-Parmer 150-mm Direct Reading Panel-Mount Flowmeters
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title