Home  >  Lab Equipment > Stirrers > Magnetic Stirrers > Compact Stirrers > Cole-Parmer Battery-Powered Magnetic Stirrer
Cole-Parmer Battery-Powered Magnetic Stirrer
Cole-Parmer Battery-Powered Magnetic Stirrer
  • Portable—ideal for field analysis
  • Stepless speed control to 700 rpm
Take this compact, battery-powered magnetic stirrer anywhere you need vigorous sample
stirring. Suitable for use inside incubators, fume hoods, or glove boxes. Or, use it in the lab and eliminate the need for cumbersome extension cords. Stirrer features stepless speed control knob, corrosion-resistant epoxy-coated metal case, nonskid rubber top plate, and four rubber feet.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer Battery Powered Magnetic Stirrer
Cole-Parmer Battery-Powered Magnetic Stirrer
Product Type Stirrer (Magnetic),Max load capacity lbs 10,Speed (rpm) low 200
EW-04804-01
In stock.
$291.00 USD/EACH
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Lab Equipment > Stirrers > Magnetic Stirrers > Compact Stirrers > Cole-Parmer Battery-Powered Magnetic Stirrer
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title