log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Gear Pumps > Gear Pump Systems/Skids > Cole-Parmer Benchtop Digital Gear Pump Drive Systems
Cole-Parmer Benchtop Digital Gear Pump Drive Systems
Cole-Parmer Benchtop Digital Gear Pump Drive Systems
Calibrate, copy, and dispense features produce precise, pulseless batching and dispensing. Displacement and flow values for all Micropump A-mount pump heads are stored in memory—simply enter the pump head size (gear code) to retrieve the displacement per revolution. IP23-rated enclosure protects the internal components.
Controller has a DB15F connection to the optional foot switch for hands-free on/off control (order foot switch separately to the right).
Service kits 07144-42, 74012-02, -42, and -52 include: gears, suction shoe, shaft seals, and
springs.
Service kits 74012-12, -22, and -32 include: gears, bushings, and seals.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 017 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.017 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-70
Usually ships in 9 days.
$2,188.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 1 17 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 1.17 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-40
Call 1-888-358-4705 for availability.
$2,325.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 316 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.316 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-10
Call 1-888-358-4705 for availability.
$2,104.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 261 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.261 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-50
Call 1-888-358-4705 for availability.
$2,174.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 91 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.91 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-30
Usually ships in 7 days.
$2,104.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 261 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.261 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-55
Call 1-888-358-4705 for availability.
$2,209.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 64 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.64 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-25
Call 1-888-358-4705 for availability.
$2,138.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 91 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.91 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-35
Call 1-888-358-4705 for availability.
$2,138.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 64 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.64 mL/rev, 115 VAC
Max temperature 104F (40C),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-20
Call 1-888-358-4705 for availability.
$2,104.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 017 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.017 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-75
Call 1-888-358-4705 for availability.
$2,223.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 092 mL rev 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.092 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-00
Call 1-888-358-4705 for availability.
$2,188.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 316 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.316 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-15
Call 1-888-358-4705 for availability.
$2,138.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 1 17 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 1.17 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-45
Call 1-888-358-4705 for availability.
$2,359.00 USD/EACH
Qty:
Representative photo only Cole Parmer Gear Pump System Benchtop Digital Drive 0 092 mL rev 220 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.092 mL/rev, 220 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-65
Call 1-888-358-4705 for availability.
$1,844.00 USD/EACH
Qty:
EW-74014-60 Cole-Parmer<small><sup>&reg;</sup></small> Gear Pump System, Benchtop Digital Drive, 0.092 mL/rev, 115 VAC
Cole-Parmer Gear Pump System, Benchtop Digital Drive, 0.092 mL/rev, 115 VAC
Max temperature 104F (40C) (system),Duty cycle Continuous,Viscosity 0.2 to 1500 cp
EW-74014-60
Call 1-888-358-4705 for availability.
$1,810.00 USD/EACH
Qty:
16 results (showing 1 - 15 )
Per Page 
Home  >  Pumps > Gear Pumps > Gear Pump Systems/Skids > Cole-Parmer Benchtop Digital Gear Pump Drive Systems
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title