Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Product Type:
Price Range:
Availability:
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  pH Meters and Water Quality Products > Chloride Analyzers > Cole-Parmer Chloride Analyzer

Cole-Parmer Chloride Analyzer

Cole-Parmer Chloride Analyzer

  • More accurate and precise than any other rapid method of salt determination!
  • Measurement repeatability ensures quality assurance and control
  • Compact design saves valuable bench space
  • Chloride standards are calibrated in both mg/L and mmol/L
This automatic chloride titrator provides accurate and highly reproducible measurement results. The system is ideal for use in the food, pharmaceutical, agricultural, and petroleum industries. The system utilizes a method based on coulometric titration—an absolute method where the reagent, silver ions, is precisely and quantitatively generated at the time of analysis by passing a constant current between electrodes. The end point is detected by the use of sensing electrodes which measure the change in the solution conductivity that occurs when excess silver ions are present in the solution. Unlike a conductivity meter, this analyzer actually titrates chloride with silver as in official methods.
Automatic features provide speed and simplicity, reducing labor costs and
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer Chloride Analyzer 120v

Cole-Parmer Chloride Analyzer, 120v

Product Type Chloride Analyzer,Description Chloride analyzer
EW-02656-20
Usually ships in 6 days.
$6,892.00 USD/EACH

Qty:

Representative photo only Acid Buffer Reagent 500 ml Bottle
EW-02656-30
In stock.
$26.00 USD/EACH

Qty:

Representative photo only Silver Electrodes Anode Pack Of Three
EW-02656-34
In stock.
$109.00 USD/PACKAGE

Qty:

Representative photo only Chloride Standard 1 x 100 mL bottle

Chloride Standard, 1 x 100 mL bottle

Product Type Accessory
EW-02656-39
In stock.
$16.75 USD/EACH

Qty:

Representative photo only Silver Electrodes Two Detecting Electrodes One Cathode
EW-02656-32
In stock.
$109.00 USD/PACKAGE

Qty:

Representative photo only Electrode Cleaning Polish 25 Gram Bottle
EW-02656-40
Usually ships in 7 days.
$40.00 USD/EACH

Qty:

Representative photo only Beaker 20 ml Capacity Marked At 14 ml
EW-02656-36
Usually ships in 6 days.
$33.00 USD/EACH

Qty:

7 results (showing 1 - 7 )
Per Page 
Home  >  pH Meters and Water Quality Products > Chloride Analyzers > Cole-Parmer Chloride Analyzer

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title