United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Brand:
Cole-Parmer
(1)
Product Type:
Accessory
(1)
Price Range:
$0 to $100
(4)
$100 to $299
(2)
Availability:
In Stock
(4)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
>
Chloride Analyzers
>
Cole-Parmer Chloride Analyzer
> Cole-Parmer Chloride Analyzer Accessories
Cole-Parmer Chloride Analyzer Accessories
More accurate and precise than any other rapid method of salt determination!
Measurement repeatability ensures quality assurance and control
Compact design saves valuable bench space
Chloride standards are calibrated in both mg/L and mmol/L
This automatic chloride titrator provides accurate and highly reproducible measurement results. The system is ideal for use in the food, pharmaceutical, agricultural, and petroleum industries. The system utilizes a method based on coulometric titrationan absolute method where the reagent, silver ions, is precisely and quantitatively generated at the time of analysis by passing a constant current between electrodes. The end point is detected by the use of sensing electrodes which measure the change in the solution conductivity that occurs when excess silver ions are present in the solution. Unlike a conductivity meter, this analyzer actually titrates chloride with silver as in official methods.
Automatic features provide speed and simplicity, reducing labor costs and
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Acid Buffer Reagent, 500 ml Bottle
EW-02656-30
In stock.
$26.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Chloride Analyzer Accessories
Silver Electrodes, Anode. Pack Of Three
EW-02656-34
In stock.
$109.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Chloride Analyzer Accessories
Chloride Standard, 1 x 100 mL bottle
Product Type Accessory
EW-02656-39
In stock.
$16.75 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Chloride Analyzer Accessories
Silver Electrodes, Two Detecting Electrodes, One Cathode
EW-02656-32
In stock.
$109.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Chloride Analyzer Accessories
Electrode Cleaning Polish, 25 Gram Bottle
EW-02656-40
Usually ships in 7 days.
$40.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Chloride Analyzer Accessories
Beaker, 20 ml Capacity, Marked At 14 ml
EW-02656-36
Usually ships in 6 days.
$33.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Chloride Analyzer Accessories
6
results (showing
1
-
6
)
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
>
Chloride Analyzers
>
Cole-Parmer Chloride Analyzer
> Cole-Parmer Chloride Analyzer Accessories
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title