log in or register
Brand:
Product Type:
Price Range:
Availability:
Max pressure (psi):
100 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pressure & Vacuum Instruments > Switches > Cole-Parmer Compact Pressure/Vacuum Switches
Cole-Parmer Compact Pressure/Vacuum Switches
Cole-Parmer Compact Pressure/Vacuum Switches
  • Compact size is ideal for tight areas
Use these switches to control pumps or valves or to alert the operator when the pressure/vacuum increases or decreases. Switches feature a SPDT relay that is actuated when the pressure/vacuum setpoint is exceeded. Easily adjust switches to the desired setpoint. Simply loosen the knurled locking ring, set the sliding gauge pointer to the desired setting, and tighten the locking ring—the switch is ready to use. Includes mounting bracket and 11"L wire leads.
 Product Detail
Sort
 Availability
 Price
Representative photo only Pressure Switch 3 0 to 96 5 psig decreasing ranges 4 5 to 100 0 psig increasing range 100 psig maximum system pressure greater than 20 CPM
Pressure Switch; 3.0 to 96.5 psig decreasing ranges, 4.5 to 100.0 psig increasing range; 100 psig maximum system pressure; greater than 20 CPM
Product Type Vacuum gauges/controllers,Output SPDT relay rated for 250 VAC/28 VDC, 5 A,Max pressure psi 100
EW-68061-10
In stock.
$91.00 USD/EACH
Qty:
Representative photo only Vacuum Switch 1 6 to 27 0 Hg decreasing ranges 2 7 to 28 2 Hg increasing range 29 9 Hg maximum system pressure greater than 60 CPM
Vacuum Switch; 1.6 to 27.0 " Hg decreasing ranges, 2.7 to 28.2 " Hg increasing range; 29.9" Hg maximum system pressure; greater than 60 CPM
Product Type Pressure/vacuum switches,Output SPDT relay rated for 250 VAC/28 VDC, 5 A,Brand Cole-Parmer
EW-68061-00
Usually ships in 8 days.
$91.00 USD/EACH
Qty:
Representative photo only Pressure Switch 0 8 to 28 5 psig decreasing ranges 1 6 to 30 0 psig increasing range 30 psig maximum system pressure greater than 60 CPM
Pressure Switch; 0.8 to 28.5 psig decreasing ranges, 1.6 to 30.0 psig increasing range; 30 psig maximum system pressure; greater than 60 CPM
Product Type Vacuum gauges/controllers,Output SPDT relay rated for 250 VAC/28 VDC, 5 A,Brand Cole-Parmer
EW-68061-06
Usually ships in 15 days.
$91.00 USD/EACH
Qty:
EW-68061-08 Pressure Switch; 2.0 to 48.0 psig decreasing ranges, 3.0 to 50.0 psig increasing range; 50 psig maximum system pressure; greater than 60 CPM
Pressure Switch; 2.0 to 48.0 psig decreasing ranges, 3.0 to 50.0 psig increasing range; 50 psig maximum system pressure; greater than 60 CPM
Product Type Vacuum gauges/controllers,Output SPDT relay rated for 250 VAC/28 VDC, 5 A,Brand Cole-Parmer
EW-68061-08
Usually ships in 12 days.
$91.00 USD/EACH
Qty:
Representative photo only Pressure Switch 15 0 to 485 0 psig decreasing ranges 20 0 to 500 0 psig increasing range 500 psig maximum system pressure greater than 20 CPM
Pressure Switch; 15.0 to 485.0 psig decreasing ranges, 20.0 to 500.0 psig increasing range; 500 psig maximum system pressure; greater than 20 CPM
Product Type Vacuum gauges/controllers,Output SPDT relay rated for 250 VAC/28 VDC, 5 A,Brand Cole-Parmer
EW-68061-14
Call 1-888-358-4705 for availability.
$91.00 USD/EACH
Qty:
Representative photo only Pressure Switch 30 00 to 150 0 psig max pressure 600 psig
Pressure Switch, 30.00 to 150.0 psig, max pressure 600 psig
Product Type Vacuum gauges/controllers,Model P88G-5-C15TS
EW-68060-08
Call 1-888-358-4705 for availability.
$45.00 USD/EACH
Qty:
Representative photo only Compound Switch 28 0 Hg to 3 5 psig max pressure 29 9 Hg 600 psig
Compound Switch, 28.0" Hg to 3.5 psig, max pressure 29.9" Hg (600 psig)
Product Type Vacuum gauges/controllers,Brand Whitman Controls
EW-68060-02
Usually ships in 7 days.
$53.00 USD/EACH
Qty:
Representative photo only Pressure Switch 1 5 to 3 5 psig max pressure 600 psig
Pressure Switch, 1.5 to 3.5 psig, max pressure 600 psig
Product Type Vacuum gauges/controllers
EW-68060-04
Usually ships in 15 days.
$39.00 USD/EACH
Qty:
Representative photo only Pressure Switch 7 5 to 242 5 psig decreasing ranges 9 7 to 250 0 psig increasing range 250 psig maximum system pressure greater than 20 CPM
Pressure Switch; 7.5 to 242.5 psig decreasing ranges, 9.7 to 250.0 psig increasing range; 250 psig maximum system pressure; greater than 20 CPM
Product Type Vacuum gauges/controllers,Output SPDT relay rated for 250 VAC/28 VDC, 5 A,Brand Cole-Parmer
EW-68061-12
Usually ships in 12 days.
$91.00 USD/EACH
Qty:
Representative photo only Pressure Switch 100 0 to 500 0 psig max pressure 600 psig
Pressure Switch, 100.0 to 500.0 psig, max pressure 600 psig
Product Type Vacuum gauges/controllers
EW-68060-10
Usually ships in 14 days.
$30.00 USD/EACH
Qty:
Representative photo only Vacuum Switch 6 0 to 28 0 Hg max pressure 29 9 Hg
Vacuum Switch, 6.0 to 28.0" Hg, max pressure 29.9" Hg
Product Type Pressure/vacuum switches
EW-68060-00
Call 1-888-358-4705 for availability.
$39.00 USD/EACH
Qty:
11 results (showing 1 - 11 )
Per Page 
Home  >  Pressure & Vacuum Instruments > Switches > Cole-Parmer Compact Pressure/Vacuum Switches
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title