United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$1,500 to $1,700
(1)
$1,700 to $1,999
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Gear Pumps
>
Gear Pump Drives - For Micropump and Other Pump Heads
> Cole-Parmer Console Digital Dispensing Drive with Remote Control for Micropump A Mount Gear Pumps
Cole-Parmer Console Digital Dispensing Drive with Remote Control for Micropump A Mount Gear Pumps
Easily operate this console drive from the front panel or use the optional foot switch (sold separately) for hands-free on/off control. The IP23-rated enclosure protects internal components from damage. Digital optical encoder feedback signal keeps speed control to ±0.3%.
What's included:
the 115 VAC model has a 6-ft modular line cord with U.S. standard plug. The 230 VAC model has a 6-ft modular line cord with IEC 320/CEE 22 connectorplease specify country to obtain correct plug when ordering.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Cole-Parmer Console Digital Dispensing Drive with Remote Control for Micropump A-mount Pump Heads, 115 VAC
Duty cycle Continuous,Motor type Permanent magnet DC (PMDC), IP23,Motor hp 1/10
EW-75211-30
In stock.
$1,688.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Console Digital Dispensing Drive
Cole-Parmer Console Digital Dispensing Drive with Remote Control for Micropump A-Mount Pump Heads, 230 VAC
Duty cycle continuous,Motor type Permanent magnet DC (PMDC), IP23,Motor hp 1/10
EW-75211-35
In stock.
$1,725.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Console Digital Dispensing Drive with Remote Control for Micropump A Mount Gear Pumps
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Gear Pumps
>
Gear Pump Drives - For Micropump and Other Pump Heads
> Cole-Parmer Console Digital Dispensing Drive with Remote Control for Micropump A Mount Gear Pumps
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title