United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pressure & Vacuum Instruments
>
Vacuum Controllers, Gauges, and Systems
> Cole-Parmer Handheld Digital Vacuum Gauge
Cole-Parmer Handheld Digital Vacuum Gauge
Use optional extension cables to take remote pressure measurements
Our compact, lightweight gauge is ideal for vacuum system use--measures vacuum from 0 to 1500 mbar. Durable ceramic sensor is enclosed in polyethyletherketone (PEEK) housing to offer excellent chemical compatibility and provide years of reliable service. Sensor is detachable--order optional extension cables to take remote readings up to 16 feet (5 meters) from gauge.
The ABS plastic housing resists many chemicals. Rear-mounted clip makes it easy to carry or mount the gauge. Vacuum gauge comes with rechargeable NiCd battery, 115 VAC adapter/battery recharger, and hose barb adapter. Order optional adapters to use gauge with flanged or glass connections.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Handheld Vacuum Meter, 0-1999 mbar
Product Type Vacuum gauges/controllers,CE Compliance Yes,Display 3.5-digit LCD
EW-68980-02
Call 1-888-358-4705 for availability.
$679.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Handheld Digital Vacuum Gauge
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Pressure & Vacuum Instruments
>
Vacuum Controllers, Gauges, and Systems
> Cole-Parmer Handheld Digital Vacuum Gauge
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title