log in or register
Availability:
Max pressure (psi):
100 (1)
30 (1)
15 (1)
1000 (1)
500 (1)
4000 (1)
10,000 (1)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pressure & Vacuum Instruments > Gauges > Digital Pressure Gauges and Meters > Cole-Parmer Digital Gauge with 0.25% Accuracy > Cole-Parmer High-Accuracy Digital Gauge with 4-digit LCD and peak hold
Cole-Parmer High-Accuracy Digital Gauge with 4-digit LCD and peak hold
Cole-Parmer High-Accuracy Digital Gauge with 4-digit LCD and peak hold
  • Injection-molded case and 316 stainless steel wetted parts resist rust and corrosion
Gauges offer high accuracy plus the convenience of a digital display. The semiconductor sensing element measures pressure with ±0.25% accuracy and lasts for millions of cycles. Adjust zero and span potentiometers on front of case.
Models 68920-00 through -20 have a 31/2-digit LCD and approximately 500 hours of battery life. Models 68920-30 through -74 have a four-digit LCD, peak hold function, TARE function to rezero the gauge, 16-minute auto-off function, and approximately 1000 hours of battery life.
All models feature automatic shut-off operation to help prolong battery life.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 5000 PSI 4 Digit LCD
Cole-Parmer High-Accuracy Digital Gauge, 0 to 5000 PSI, 4-Digit LCD
Product Type Digital gauges,Max pressure psi 4000,Dial size " Dia 2.5
EW-68920-44
Call 1-888-358-4705 for availability.
$415.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 100 0 psig 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 100.0 psig, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Max pressure psi 100,Dial size " Dia 2.5
EW-68920-38
In stock.
$415.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 30 0 psig 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 30.0 psig, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Max pressure psi 30,Dial size " Dia 2.5
EW-68920-34
In stock.
$408.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 1000 mbar 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 1000 mbar, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Dial size " Dia 2.5,Display 4-digit LCD, 0.6" H
EW-68920-50
In stock.
$408.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 15 00 psig 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 15.00 psig, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Max pressure psi 15,Dial size " Dia 2.5
EW-68920-32
Call 1-888-358-4705 for availability.
$408.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 10 000 psig 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 10,000 psig, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Max pressure psi 10,000,Dial size " Dia 2.5
EW-68920-48
Call 1-888-358-4705 for availability.
$415.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 500 psig 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 500 psig, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Max pressure psi 500,Dial size " Dia 2.5
EW-68920-40
Call 1-888-358-4705 for availability.
$415.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 700 kPa 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 700 kPa, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Dial size " Dia 2.5,Display 4-digit LCD, 0.6" H
EW-68920-58
Call 1-888-358-4705 for availability.
$415.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 1000 psig 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 1000 psig, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Max pressure psi 1000,Dial size " Dia 2.5
EW-68920-42
Call 1-888-358-4705 for availability.
$424.00 USD/EACH
Qty:
Representative photo only Cole Parmer High Accuracy Digital Gauge 0 to 40 00 kg cm2 4 Digit LCD Peak Hold
Cole-Parmer High-Accuracy Digital Gauge, 0 to 40.00 kg/cm2, 4-Digit LCD, Peak Hold
Product Type Digital gauges,Dial size " Dia 2.5,Display 4-digit LCD, 0.6" H
EW-68920-70
In stock.
$415.00 USD/EACH
Qty:
10 results (showing 1 - 10 )
Per Page 
Home  >  Pressure & Vacuum Instruments > Gauges > Digital Pressure Gauges and Meters > Cole-Parmer Digital Gauge with 0.25% Accuracy > Cole-Parmer High-Accuracy Digital Gauge with 4-digit LCD and peak hold
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title