Home  >  Laboratory Supplies & Consumables > Film and Foil > Cole-Parmer PTFE Film
Cole-Parmer PTFE Film
Cole-Parmer PTFE Film
Chemical resistant, temperature resistant, non-stick PTFE film protects benchtops, shelves, drawers, tables, trays, sinks, and other surfaces. Material is not porous, will not stain, and will not absorb odors or other contaminants. Reuseable film is easy to clean and easily cuts to size with scissors. Non-stick properties make this film ideal for use as a weighing sheet.
 Product Detail
Sort
 Availability
 Price
EW-08277-15 PTFE Film .005"thick x 12"W x 25ft rolls
PTFE Film .005"thick x 12"W x 25ft rolls
Product Type Film,Dimensions ft/roll 25,Color Opaque,Material PTFE
EW-08277-15
In stock.
$111.00 USD/ROLL
Qty:
1 results (showing 1 - 1 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Film and Foil > Cole-Parmer PTFE Film
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title