log in or register
Brand:
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Pumps > Metering Pumps > Piston Pumps > Series Q Valveless Piston Pump Systems > Cole-Parmer Precision Flow Piston Pump Systems
Cole-Parmer Precision Flow Piston Pump Systems
Cole-Parmer Precision Flow Piston Pump Systems
  • These popular drive and head combinations simplify ordering!
  • Prebuilt systems with the most popular head and drive combinations
  • Some defined systems also contain the new variable-speed controller (07104-32)
  • High repeatability and low maintenance is a key factor for these piston pump systems
Series Q pump heads have no valves for a drift-free operation—providing millions of maintenance-free cycles. There is only one moving part, resulting in accuracy better than 1%.
Flow rate adjustment could not be easier: simply move the pump head position to change the piston stroke length and, in turn, the flow rate. Flow rate can be adjusted even while the pump is operating.
Typical applications include paint, pigments, resins, instrumentation, cleaning agents, fuel cell, battery manufacturing, cosmetics, electronics manufacturing, various pilot plant configurations, food and
dairy, agricultural, and many more.
 Product Detail
Sort
 Availability
 Price
Representative photo only Piston Pump System consisting of V300 controller and RHV head 3 16 Piston Diameter 0 to 180 mL min
Piston Pump System consisting of V300 controller and RHV head, 3/16" Piston Diameter, 0 to 180 mL/min
Product Type Piston Metering Pumps,Max pressure 100 psi,Max flow rate mL/min 180
EW-07108-61
In stock.
$2,470.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S Drive adapter and Q2 head 3 8 Piston Diameter 0 to 432 mL min 90 to 130VAC
EW-07108-73
Usually ships in 10 days.
$1,663.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S Drive adapter and Q1 head 1 4 Piston Diameter 0 to 192 mL min 190 to 260VAC
EW-07108-76
Usually ships in 10 days.
$1,663.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S drive adapter and RH1 head 3 16 Piston Diameter 0 to 60 mL min 90 to 130VAC
EW-07108-71
Usually ships in 10 days.
$1,688.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S Drive adapter and RH0 head 3 16 Piston Diameter 0 to 30 mL min 190 to 260VAC
EW-07108-74
Usually ships in 10 days.
$1,688.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S Drive adapter and Q2 head 3 8 Piston Diameter 0 to 432 mL min 190 to 260VAC
EW-07108-77
Usually ships in 10 days.
$1,663.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S Drive adapter and RH0 head 3 16 Piston Diameter 0 to 30 mL min 90 to 130VAC
EW-07108-70
Usually ships in 10 days.
$1,688.00 USD/EACH
Qty:
Representative photo only Piston Pump System consisting of Masterflex L S drive adapter and RH1 head 3 16 Piston Diameter 0 to 60 mL min 190 to 260VAC
EW-07108-75
Usually ships in 10 days.
$1,688.00 USD/EACH
Qty:
8 results (showing 1 - 8 )
Per Page 
Home  >  Pumps > Metering Pumps > Piston Pumps > Series Q Valveless Piston Pump Systems > Cole-Parmer Precision Flow Piston Pump Systems
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title