log in or register
Price Range:
Availability:
Capacity (mL):
0 - 20 (2)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Laboratory Supplies & Consumables > Graduated Cylinders > Glass Graduated Cylinders > Cole-Parmer Serialized Class A Graduated Glass Cylinders
Cole-Parmer Serialized Class A Graduated Glass Cylinders
Cole-Parmer Serialized Class A Graduated Glass Cylinders
  • Available with Certificate of Traceability and Certificate of Calibration
  • Calibrated To Deliver
  • Single metric scale
  • Meet ASTM E 438
Choose cylinders with certificate of traceability to NIST standards or with certificate of calibration and traceability. All support ISO 9000 requirements and are calibrated in accordance with ASTM E 542. Cylinders meet ASTM E 694 specifications and accuracy requirements of ASTM 1272. Cylinder glass meets ASTM E 438.
Each cylinder features an easy-to-read white scale and a molded base to decrease risk of accidentally knocking it over. Models with certificate of calibration and traceability feature fused-in serial numbers.
 Product Detail
Sort
 Availability
 Price
Representative photo only Cylinder 10 ml Class A To Contain Certificate of Traceability
Cylinder, 10 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 10,Material Glass
EW-34300-17
In stock.
$75.00 USD/EACH
Qty:
Representative photo only Cylinder 1000 ml Class A To Contain Certificate of Calibration and Traceability
Cylinder, 1000 ml, Class A , To Contain, Certificate of Calibration and Traceability
Product Type Graduated Cylinder,Capacity mL 1000,Material Glass
EW-34300-30
Usually ships in 13 days or sooner.
$385.00 USD/EACH
Qty:
Representative photo only Cylinder 50 ml Class A To Deliver Certificate of Traceability
Cylinder, 50 ml, Class A , To Deliver, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 50,Material Glass
EW-34300-05
In stock.
$84.00 USD/EACH
Qty:
Representative photo only Cylinder 100 ml Class A To Contain Certificate of Traceability
Cylinder, 100 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 100,Material Glass
EW-34300-23
In stock.
$89.00 USD/EACH
Qty:
Representative photo only Cylinder 1000 ml Class A To Contain Certificate of Traceability
Cylinder, 1000 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 1000,Material Glass
EW-34300-29
In stock.
$136.00 USD/EACH
Qty:
Representative photo only Cylinder 250 ml Class A To Contain Certificate of Traceability
Cylinder, 250 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 250,Material Glass
EW-34300-25
In stock.
$94.00 USD/EACH
Qty:
EW-34300-27 Cylinder, 500 ml, Class A , To Contain, Certificate of Traceability
Cylinder, 500 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 500,Material Glass
EW-34300-27
In stock.
$120.00 USD/EACH
Qty:
Representative photo only Cylinder 100 ml Class A To Deliver Certificate of Calibration and Traceability
Cylinder, 100 ml, Class A , To Deliver, Certificate of Calibration and Traceability
Product Type Graduated Cylinder,Capacity mL 100,Material Glass
EW-34300-08
Usually ships in 9 days or sooner.
$215.00 USD/EACH
Qty:
Representative photo only Cylinder 25 ml Class A To Contain Certificate of Traceability
Cylinder, 25 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 25,Material Glass
EW-34300-19
In stock.
$85.00 USD/EACH
Qty:
Representative photo only Cylinder 250 ml Class A To Deliver Certificate of Calibration and Traceability
Cylinder, 250 ml, Class A , To Deliver, Certificate of Calibration and Traceability
Product Type Graduated Cylinder,Capacity mL 250,Material Glass
EW-34300-10
Usually ships in 14 days.
$300.00 USD/EACH
Qty:
Representative photo only Cylinder 1000 ml Class A To Deliver Certificate of Calibration and Traceability
Cylinder, 1000 ml, Class A , To Deliver, Certificate of Calibration and Traceability
Product Type Graduated Cylinder,Capacity mL 1000,Material Glass
EW-34300-16
Call 1-888-358-4705 for availability.
$299.00 USD/EACH
Qty:
Representative photo only Cylinder 500 ml Class A To Deliver Certificate of Calibration and Traceability
Cylinder, 500 ml, Class A , To Deliver, Certificate of Calibration and Traceability
Product Type Graduated Cylinder,Capacity mL 500,Material Glass
EW-34300-12
Usually ships in 9 days.
$246.00 USD/EACH
Qty:
Representative photo only Cylinder 50 ml Class A To Contain Certificate of Traceability
Cylinder, 50 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 50,Material Glass
EW-34300-21
In stock.
$89.00 USD/EACH
Qty:
Representative photo only Cylinder 2000 ml Class A To Contain Certificate of Traceability
Cylinder, 2000 ml, Class A , To Contain, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 2000,Material Glass
EW-34300-31
Usually ships in 14 days.
$241.00 USD/EACH
Qty:
Representative photo only Cylinder 250 ml Class A To Deliver Certificate of Traceability
Cylinder, 250 ml, Class A , To Deliver, Certificate of Traceability
Product Type Graduated Cylinder,Capacity mL 250,Material Glass
EW-34300-09
In stock.
$110.00 USD/EACH
Qty:
30 results (showing 1 - 15 )
Per Page 
Home  >  Laboratory Supplies & Consumables > Graduated Cylinders > Glass Graduated Cylinders > Cole-Parmer Serialized Class A Graduated Glass Cylinders
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title