log in or register
Price Range:
Availability:
Capacity (Liters):
1 to 5 (6)
CE Compliance:
Yes (6)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Lab Equipment > Baths and Circulators > Laboratory, Utility Water Baths > Cole-Parmer StableTemp Digital Utility Baths
Cole-Parmer StableTemp Digital Utility Baths
Cole-Parmer StableTemp Digital Utility Baths
  • Easy-to-set temperature control with adjustable presets for fast set point changes
  • Front panel lockout prevents unauthorized set point changes
  • Hinged lid directs coundensation back into the bath—prevents wet, messy benchtops
  • Extensive over-temperature protection
The name says it all! Our StableTemp utility baths feature unique air-jacketed heaters which provide excellent temperature specifications. The +/-0.2°C temperature uniformity and +/-0.25°C stability are ideal for heating your temperature-sensitive samples.
For optimum temperature protection, these baths feature dual thermostats—a primary, microprocessor-based temperature controller that provides +/-0.1°C precision and a secondary over temperature safety prevents samples from overheating. For optimum temperature accuracy, the primary controller can be calibrated to your reference thermometer.
The stainless steel tank is seamless—no welds to leak or rust—and the rounded corners provide an easy-to-clean interior. Hinged polycarbonate lid allows condensation to flow back.
Optional shaker platforms are available to convert the 20 and 28 L digital utility baths into shaking water baths. Allows samples to incubate and aggitate simultaneously.Laboratory, utility water baths are the ideal bath for maintaining a temperature set points over a long period of time. Different size and depth of the bath chamber to meet most every application. Units feature analog or digital temperature control with a ranges from ambient to 100° C. Options of high-temperature and low-level cutoff features to protect samples.
 Product Detail
Sort
 Availability
 Price
EW-14575-12 Water bath, 10L, 120V
Cole-Parmer StableTemp Water bath, 10L, 120V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 10
EW-14575-12
In stock.
$1,041.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 28L 120V
Cole-Parmer StableTemp Water bath, 28L, 120V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 28
EW-14575-20
Call 1-888-358-4705 for availability.
$1,533.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 28L 240V
Cole-Parmer StableTemp Water bath, 28L, 240V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 28
EW-14575-22
Call 1-888-358-4705 for availability.
$1,533.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 20L 120V
Cole-Parmer StableTemp Water bath, 20L, 120V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 20
EW-14575-16
In stock.
$1,369.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 5L 120V
Cole-Parmer StableTemp Water bath, 5L, 120V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 5
EW-14575-08
In stock.
$853.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 2L deep 120V
Cole-Parmer StableTemp Water bath, 2L deep, 120V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 2
EW-14575-00
Call 1-888-358-4705 for availability.
$701.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 10L 240V
Cole-Parmer StableTemp Water bath, 10L, 240V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 10
EW-14575-14
In stock.
$1,041.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 20L 240V
Cole-Parmer StableTemp Water bath, 20L, 240V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 20
EW-14575-18
Call 1-888-358-4705 for availability.
$1,369.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 5L 240V
Cole-Parmer StableTemp Water bath, 5L, 240V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 5
EW-14575-10
Call 1-888-358-4705 for availability.
$853.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 2L shallow 120V
Cole-Parmer StableTemp Water bath, 2L shallow, 120V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 2
EW-14575-04
Usually ships in 15 days.
$762.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 2L shallow 240V
Cole-Parmer StableTemp Water bath, 2L shallow, 240V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 2
EW-14575-06
Call 1-888-358-4705 for availability.
$762.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Water bath 2L deep 240V
Cole-Parmer StableTemp Water bath, 2L deep, 240V
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 2
EW-14575-02
Call 1-888-358-4705 for availability.
$701.00 USD/EACH
Qty:
12 results (showing 1 - 12 )
Per Page 
Home  >  Lab Equipment > Baths and Circulators > Laboratory, Utility Water Baths > Cole-Parmer StableTemp Digital Utility Baths
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title