log in or register
Price Range:
Availability:
Capacity (Liters):
1 to 5 (4)
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Lab Equipment > Baths and Circulators > Laboratory, Utility Water Baths > Cole-Parmer StableTemp Economical Water Baths
Cole-Parmer StableTemp Economical Water Baths
Cole-Parmer StableTemp Economical Water Baths
  • Our best selling utility baths offer excellent performance at affordable prices!
  • ±0.2°C stability at 37°C
  • Seamless 304 stainless steel tank and powder-coated exterior are corrosion resistant
  • Front-panel controls are safe and easy-to-use
These water baths are ideal for general-purpose applications. Bath fluids can be controlled at temperatures as high as 100°C (60° without cover) with +/-0.2° precision and uniformity. Bath temperature can be read from alcohol thermometer (included) with a range from -20 to 150°C. The steeply gabled, polycarbonate cover accommodates glassware of varying heights and tilts out of your way when loading or removing samples, allowing condensate to drain neatly back into the bath.
Laboratory, utility water baths are the ideal bath for maintaining a temperature set points over a long period of time. Different size and depth of the bath chamber to meet most every application. Units feature analog or digital temperature control with a ranges from ambient to 100° C. Options of high-temperature and low-level cutoff features to protect samples.
 Product Detail
Sort
 Availability
 Price
EW-12106-74 Economy bath, 5L, 120V, 60Hz
Cole-Parmer StableTemp Economy bath, 5L, 120V, 60Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 5
EW-12106-74
Call 1-888-358-4705 for availability.
$670.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 10L 120V 60Hz
Cole-Parmer StableTemp Economy bath, 10L, 120V, 60Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 10
EW-12106-78
In stock.
$747.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 2L 120V 60Hz
Cole-Parmer StableTemp Economy bath, 2L, 120V, 60Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 2
EW-12106-70
In stock.
$589.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 10L 240V 50Hz
Cole-Parmer StableTemp, Economy bath, 10L, 240V, 50Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 10
EW-12106-80
Call 1-888-358-4705 for availability.
$747.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 20L 120V 60Hz
Cole-Parmer StableTemp Economy bath, 20L, 120V, 60Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 20
EW-12106-82
Usually ships in 13 days.
$896.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 20L 240V 50Hz
Cole-Parmer StableTemp Economy bath, 20L, 240V, 50Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 20
EW-12106-84
Call 1-888-358-4705 for availability.
$896.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 28L 120V 60Hz
Cole-Parmer StableTemp Economy bath, 28L, 120V, 60Hz
Product Type Water Bath,Bath type Analog Water Bath,Capacity Liters 28
EW-12106-86
In stock.
$1,011.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 5L 240V 50Hz
Cole-Parmer StableTemp Economy bath, 5L, 240V, 50Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 5
EW-12106-76
Call 1-888-358-4705 for availability.
$670.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 2L 240V 50Hz
Cole-Parmer StableTemp Economy bath, 2L, 240V, 50Hz
Product Type Water Bath,Bath type Utility Water Bath,Capacity Liters 2
EW-12106-72
Call 1-888-358-4705 for availability.
$589.00 USD/EACH
Qty:
Representative photo only Cole Parmer StableTemp Economy bath 28L 240V 50Hz
Cole-Parmer StableTemp Economy bath, 28L, 240V, 50Hz
Product Type Water Bath,Bath type Analog Water Bath,Capacity Liters 28
EW-12106-88
Usually ships in 15 days.
$1,011.00 USD/EACH
Qty:
10 results (showing 1 - 10 )
Per Page 
Home  >  Lab Equipment > Baths and Circulators > Laboratory, Utility Water Baths > Cole-Parmer StableTemp Economical Water Baths
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title