log in or register
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Lab Equipment > Ultrasonic Cleaners > Standard Ultrasonic Cleaners > Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater, Digital Timer, Temperature Monitor, and Degas Feature
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater, Digital Timer, Temperature Monitor, and Degas Feature
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater, Digital Timer, Temperature Monitor, and Degas Feature
  • Aggressive cleaning ability in a variety of sizes
  • Sturdy, noncorrosive units handle all types of cleaning tasks
  • Sweep frequency ensures uniform cleaning activity
Operating at 40 kHz, these ultrasonic cleaners provide aggressive cleaning power. Easy-to-clean, corrosion-resistant 304 stainless steel tanks have no corner welds or other dirt-catching surfaces. The units are enclosed in durable, powder-coated steel skirts.
Cleaners with heater, digital timer, temperature monitor, and degas feature are perfect for your critical cleaning applications. The degas feature eliminates air bubbles from the solution for more efficient cleaning. Set degas function up to 99 minutes. Adjustable heater sets tank temperatures from 34 to 156°F (1 to 69°C). Cleaner monitors solution temperature from 50 to 167°F (10 to 75°C). Over-temperature protection prevents overheating; cleaner automatically shuts down when
 Product Detail
Sort
 Availability
 Price
EW-08895-16 Cole-Parmer<small><sup>&reg;</sup></small> Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 gal, 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1,Power VAC 115
EW-08895-16
In stock.
$775.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 3 4 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 3/4 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 115
EW-08895-04
In stock.
$580.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 2 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 2 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 115
EW-08895-40
Call 1-888-358-4705 for availability.
$1,430.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 5 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 5 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 5.5,Power VAC 115
EW-08895-64
Call 1-888-358-4705 for availability.
$2,500.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 1 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1.5,Power VAC 115
EW-08895-28
Call 1-888-358-4705 for availability.
$1,120.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 1 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1,Power VAC 220
EW-08895-22
Call 1-888-358-4705 for availability.
$925.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 2 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 2 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 220
EW-08895-46
Call 1-888-358-4705 for availability.
$1,730.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 1 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 1 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1.5,Power VAC 220
EW-08895-34
Call 1-888-358-4705 for availability.
$1,450.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 3 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 3 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 3.5,Power VAC 220
EW-08895-58
Call 1-888-358-4705 for availability.
$2,200.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 3 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 3 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 3.5,Power VAC 115
EW-08895-52
Call 1-888-358-4705 for availability.
$1,790.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 5 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 5 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 5.5,Power VAC 220
EW-08895-70
Call 1-888-358-4705 for availability.
$3,240.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer 3 4 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Digital Timer, 3/4 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 220
EW-08895-10
Call 1-888-358-4705 for availability.
$675.00 USD/EACH
Qty:
12 results (showing 1 - 12 )
Per Page 
Home  >  Lab Equipment > Ultrasonic Cleaners > Standard Ultrasonic Cleaners > Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater, Digital Timer, Temperature Monitor, and Degas Feature
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title