Home  >  Lab Equipment > Ultrasonic Cleaners > Standard Ultrasonic Cleaners > Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer
  • Aggressive cleaning ability in a variety of sizes
  • Sturdy, noncorrosive units handle all types of cleaning tasks
  • Sweep frequency ensures uniform cleaning activity
Operating at 40 kHz, these ultrasonic cleaners provide aggressive cleaning power. Easy-to-clean, corrosion-resistant 304 stainless steel tanks have no corner welds or other dirt-catching surfaces. The units are enclosed in durable, powder-coated steel skirts.
Cleaners with heater and mechanical timer offer greater cleaning capability—ideal for heat cleaning any size parts. Maximum heater temperature is 140°F (60°C). Set the cleaning cycle up to 60 minutes or use the hold function for continuous operation.
What's included: stainless steel lid, nonskid feet, and a 7-1⁄2 ft power cord. The 1, 1-1⁄2, 2-1⁄2, 3-1⁄2, and 5-1⁄2 gallon cleaners have a bottom-mounted drain and include tank drain kits.
 Product Detail
Sort
 Availability
 Price
EW-08895-38 Cole-Parmer<small><sup>&reg;</sup></small> Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 2 1/2 gal, 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 2 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 115
EW-08895-38
Call 1-888-358-4705 for availability.
$1,293.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 3.5,Power VAC 115
EW-08895-50
Call 1-888-358-4705 for availability.
$1,613.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 4 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3/4 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 220
EW-08895-08
Call 1-888-358-4705 for availability.
$613.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 4 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3/4 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 0.75,Power VAC 115
EW-08895-02
In stock.
$484.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 1 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 1 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1,Power VAC 115
EW-08895-14
In stock.
$685.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 1 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 1 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1.5,Power VAC 115
EW-08895-26
Call 1-888-358-4705 for availability.
$980.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 1 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 1 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1.5,Power VAC 220
EW-08895-32
Call 1-888-358-4705 for availability.
$1,203.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 3.5,Power VAC 220
EW-08895-56
Call 1-888-358-4705 for availability.
$2,040.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 2 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 2 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 2.5,Power VAC 220
EW-08895-44
Call 1-888-358-4705 for availability.
$1,640.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 1 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 1 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 1,Power VAC 220
EW-08895-20
Call 1-888-358-4705 for availability.
$856.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 5 1 2 gal 220 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 5 1/2 gal, 220 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 5.5,Power VAC 220
EW-08895-68
Call 1-888-358-4705 for availability.
$2,810.00 USD/EACH
Qty:
Representative photo only Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 5 1 2 gal 115 VAC
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 5 1/2 gal, 115 VAC
Product Type Ultrasonic Cleaner,Capacity gal. 5.5,Power VAC 115
EW-08895-62
Call 1-888-358-4705 for availability.
$2,230.00 USD/EACH
Qty:
12 results (showing 1 - 12 )
Per Page 
Home  >  Lab Equipment > Ultrasonic Cleaners > Standard Ultrasonic Cleaners > Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title