United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$0 to $100
(1)
$100 to $299
(1)
$300 to $499
(1)
Availability:
In Stock
(1)
Volume (Liters):
0 to 3
(2)
10 to 20
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Laboratory Cleaning Supplies
>
Cleaning Solutions
>
Surface Cleaners
> Cole-Parmer Surface-Cleanse/930
Cole-Parmer Surface-Cleanse/930
Leaves no charged residue—perfect for cleaning electrical components!
Safely wash away grease, petroleum, fingerprints, oils, scale, oxides, and metallic residues from glass, aluminum, nickel, lead, zinc, ceramic, and plastic without surface damages typically caused by acidic or alkaline cleaners. Use to clean filter membranes and other delicate items.
Non-flammable and non-hazardous under U.S. federal regulations for shipping, storage, and disposal. Meets USDA guidelines for incidental contact with food processing systems.
Contains no CFCs, phosphates, silicates, borates, halogens, or phenols.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Surface-Cleanse/930 Liquid Concentrate, 20 L
Product Type Equipment Cleaner,Format Liquid,Volume Liters 20
EW-18103-30
Usually ships in 9 days.
$360.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Surface-Cleanse/930
Surface-Cleanse/930 Liquid Concentrate, 1L
Product Type Equipment Cleaner,Format Liquid,Volume Liters 1
EW-18103-00
In stock.
$33.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer
Surface-Cleanse/930
Surface-Cleanse/930 Liquid Concentrate, 1L, 12 per case
Product Type Equipment Cleaner,Format Liquid,Volume Liters 1
EW-18103-10
Usually ships in 6 days.
$270.00 USD/CASE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cole-Parmer Surface-Cleanse/930
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Laboratory Cleaning Supplies
>
Cleaning Solutions
>
Surface Cleaners
> Cole-Parmer Surface-Cleanse/930
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title