Colorimeters
Colorimeters
A colorimeter uses a light source of a particular wavelength, a color-changing, colorimetry reagent, and a light detector to determine the concentration of a particular substance, usually in an aqueous or water-based sample. After mixing the sample with a specific color-changing reagent, the sample is place within the sample chamber of the colorimeter, where light of a specific wavelength is passed through it. The desired substance’s concentration measurement will react with the reagent to change the color of the sample. The concentration of the parameter is proportionate to color change intensity. Since different chemical substances absorb different visual light frequencies, the detector can determine the absorbance of the light passed through the sample based on the intensity of the sample color. A colorimeter is commonly used to test the concentration of disinfectants like chlorine used in drinking water, pool and spa applications, industrial wastewater plants, paper manufacturing, and other industries. A colorimeter can be a single-parameter, multi-parameter. The colorimeter can vary the visible light wavelength to measure a variety of parameters like fluoride, nitrate, nitrite, sulfate, phosphorous and others.
 Product Detail
Sort
 Availability
 Price
Representative photo only Hach DR 870 Colorimeter
Hach DR/870 Colorimeter
Product Type Color/Gloss Meters,Display LCD,Waterproof Yes
EW-99573-00
In stock.
$1,024.00 USD/EACH
Qty:
Representative photo only Oakton C 301 colorimeter pH free and total chlorine
Oakton C-301 colorimeter; pH, free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-30
In stock.
$426.00 USD/EACH
Qty:
EW-35645-20 Oakton<small><sup>&reg;</sup></small> C-201 colorimeter; free and total chlorine.
Oakton C-201 colorimeter; free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-20
In stock.
$386.00 USD/EACH
Qty:
Representative photo only LaMotte Single Test Colorimeter Kit Chlorine free and total in liquid form
LaMotte Single Test Colorimeter Kit, Chlorine free and total in liquid form
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-99562-12
Usually ships in 5 days.
$369.00 USD/EACH
Qty:
EW-99599-00 Orbeco-Hellige MC500<small><sup>&#153;</sup></small> multiparameter colorimeter
Orbeco-Hellige MC500 multiparameter colorimeter
Product Type Color/Gloss Meters,Display graphic display,Output IR interface for data transfer, RJ45 connector for internet updates
EW-99599-00
Usually ships in 7 days.
$1,050.00 USD/EACH
Qty:
EW-99561-30 HF scientific Chlorine Pocket Colorimeter
HF scientific Chlorine Pocket Colorimeter
Product Type Color/Gloss Meters,Display LCD,Waterproof Yes
EW-99561-30
Usually ships in 13 days.
$399.00 USD/EACH
Qty:
EW-99565-08 LaMotte Multiwavelength Colorimeter, 100 to 240 VAC
LaMotte Multiwavelength Colorimeter, 100 to 240 VAC
Product Type Color/Gloss Meters,Display LCD,Output USB
EW-99565-08
Usually ships in 7 days.
$999.00 USD/EACH
Qty:
Representative photo only Oakton C 105 colorimeter for ozone
Oakton C-105 colorimeter; for ozone
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-15
Call 1-888-358-4705 for availability.
$337.00 USD/EACH
Qty:
EW-99562-10 LaMotte Single Test Colorimeter
LaMotte Single Test Colorimeter Kit; Chlorine free and total in tablet form
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-99562-10
Usually ships in 8 days.
$369.00 USD/EACH
Qty:
EW-99570-05 THERMO AQ3070 FREE AND TOTAL CHLORINE COLORIMETER
Thermo Scientific AQ3070 Free and Total Chlorine Colorimeter
Product Type Single wavelength colorimeter,Display LED,CE Compliance Yes
EW-99570-05
Usually ships in 8 days or sooner.
$386.00 USD/EACH
Qty:
See all
Representative photo only Oakton C 401 colorimeter pH cyanuric acid free and total chlorine
Oakton C-401 colorimeter; pH, cyanuric acid, free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-40
Call 1-888-358-4705 for availability.
$452.00 USD/EACH
Qty:
Representative photo only Orbeco Hellige SC400 Colorimeter Manganese LR
Orbeco-Hellige SC400 Colorimeter, Manganese LR
Product Type Color/Gloss Meters,Display LCD,Waterproof Yes
EW-99561-58
Usually ships in 5 days.
$385.00 USD/EACH
Qty:
Representative photo only Oakton C 103 colorimeter for chlorine dioxide
Oakton C-103 colorimeter; for chlorine dioxide
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-13
Call 1-888-358-4705 for availability.
$340.00 USD/EACH
Qty:
EW-99561-76 SC450 COLORIMETER
Orbeco-Hellige SC450 Colorimeter, Free and Total Chlorine Kit
Product Type Single Wavelength Colorimeter,CE Compliance Yes,Datalogging 16 data sets
EW-99561-76
Usually ships in 6 days.
$379.00 USD/EACH
Qty:
See all
EW-99561-89 SC450 COLORIMETER
Orbeco-Hellige SC450 Colorimeter, Manganese (high) Kit
Product Type Single Wavelength Colorimeter,CE Compliance Yes,Datalogging 16 data sets
EW-99561-89
Usually ships in 6 days.
$394.00 USD/EACH
Qty:
See all
49 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title