United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Colorimetry Reagents
(88)
Colorimeters
(49)
Brand:
Hach
(21)
Orbeco-Hellige
(20)
DPD Powder Pop
(15)
[+]
Show more
(10)
LaMotte
(14)
Oakton
(10)
THERMO SCIENTIFIC
(8)
HF SCIENTIFIC
(6)
Thermo Scientific
(3)
YSI
(1)
Thermo Scientific HAAKE
(1)
HF Scientific
(1)
ORBECO HELLIGE
(1)
HF scientific
(1)
[-]
Show less
Product Type:
Water Testing Kit
(76)
Color/Gloss Meters
(20)
Single Wavelength Colorimeter
(12)
[+]
Show more
(8)
Fluorimeter
(6)
Accessory
(5)
Powder reagent
(2)
Primary Chlorine Standard
(1)
Colorimetry Accessories
(1)
Single wavelength colorimeter
(1)
Colorimeter
(1)
Standard
(1)
[-]
Show less
Price Range:
$0 to $100
(75)
$100 to $299
(20)
$300 to $499
(36)
[+]
Show more
(3)
$500 to $799
(1)
$800 to $1,099
(4)
$2,000 and Over
(1)
[-]
Show less
Availability:
In Stock
(37)
CE Compliance:
Yes
(21)
Waterproof:
Yes
(15)
Datalogging:
16 data sets
(13)
Yes
(12)
Output:
USB
(1)
IR interface for data transfer, RJ45 connector for internet updates
(1)
Learn more:
Bitter Brew
[-]
Show Less
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
pH Meters and Water Quality Products
> Colorimetry
Colorimetry
Choose from our comprehensive line of Colorimetry and other pH Meters and Water Quality Products products.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Hach DR/870 Colorimeter
Product Type Color/Gloss Meters,Display LCD,Waterproof Yes
EW-99573-00
In stock.
$1,024.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hach DR/870 Colorimeter
DPD Total Chlorine Reagent Powder Pillow Packs, 25 mL, 1000 Tests
Product Type Water Testing Kit,Model RP013-3
EW-99560-61
Usually ships in 7 days.
$180.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Powder Pack and Tube Test Reagents
Oakton C-301 colorimeter; pH, free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-30
In stock.
$426.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Colorimeter Kits
DPD Powder Pop Dispenser for Free Chlorine, 1000 Tests, 10 mL
Product Type Water Testing Kit,Brand DPD Powder Pop,Model 10504C
EW-99560-16
In stock.
$155.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
DPD Powder Pop Dispensers
Hach Total Chlorine (USEPA) Test Kit
Product Type Water Testing Kit,Brand Hach,Model 21056-69
EW-99574-05
In stock.
$21.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hach
Colorimetry Test Kits
Oakton C-201 colorimeter; free and total chlorine.
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-35645-20
In stock.
$386.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton
Colorimeter Kits
DPD Powder Pop Dispenser for Free Chlorine, 1000 Tests, 5 mL
Product Type Water Testing Kit,Brand DPD Powder Pop,Model 10500C
EW-99560-12
In stock.
$135.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
DPD Powder Pop Dispensers
DPD Free Chlorine Reagent Powder Pillow Packs, 10 mL, 100 tests
Product Type Water Testing Kit,Model RP010-0
EW-99560-54
In stock.
$19.90 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Powder Pack and Tube Test Reagents
Replacement free chlorine test kit, for Oakton colorimeters, 100 tests/kit.
Product Type Accessory,Brand Oakton
EW-35645-64
In stock.
$25.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Replacement Colorimeter Reagents
Hach Free Chlorine (USEPA) Test Kit
Product Type Water Testing Kit,Brand Hach,Model 21055-69
EW-99574-04
In stock.
$21.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Hach
Colorimetry Test Kits
Replacement total chlorine test kit, for Oakton colorimeters, 100 tests/kit.
Product Type Accessory,Brand Oakton
EW-35645-66
In stock.
$25.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Replacement Colorimeter Reagents
LaMotte Single Test Colorimeter Kit, Chlorine free and total in liquid form
Product Type Color/Gloss Meters,Display LCD,CE Compliance Yes
EW-99562-12
Usually ships in 5 days.
$369.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
LaMotte Single Test Colorimeter Kits
Orbeco-Hellige MC500
multiparameter colorimeter
Product Type Color/Gloss Meters,Display graphic display,Output IR interface for data transfer, RJ45 connector for internet updates
EW-99599-00
Usually ships in 7 days.
$1,050.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Orbeco-Hellige MC500 Multiparameter Colorimeter
Oakton Secondary Standards Chlorine Kit
Product Type Standard,Brand Oakton
EW-35645-70
In stock.
$86.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Oakton Replacement Colorimeter Reagents
DPD Powder Pop Dispenser for Total Chlorine, 1000 Tests, 5 mL
Product Type Water Testing Kit,Brand DPD Powder Pop,Model 10502C
EW-99560-14
In stock.
$135.00 USD/PACKAGE
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
DPD Powder Pop Dispensers
137
results (showing
1
-
15
)
1
2
3
4
5
6
Next >
>>
Per Page
5
10
15
25
50
Home
>
pH Meters and Water Quality Products
> Colorimetry
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title