United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Positive Displacement Pumps
>
Diaphragm Pumps
>
All-Plastic Double-Diaphragm Pumps
> Continuous-Duty Pumps
Continuous-Duty Pumps
No metal wetted parts permit use with corrosive liquids
All models feature factory-set pressure limits to protect pumps from damagepumping stops when high limit is reached, resumes when pressure decreases to low setting.
Multi-chambered pump offers efficient transfer of mild acids, bases, and light oils. Thermal overload protected, permanent-magnet motor. Pump includes 6-ft power cord with three-prong plug. Order replacement diaphragms or valve kits under accessories.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
All-plastic, continous-duty double-diaphragm pump, polypropylene body/Santoprene diaphragm, 3.5 GPM, 115 VAC
Product Type Diaphragm Pumps,Max flow rate LPM 13.2,Max pressure 40 psi
EW-75102-00
Call 1-888-358-4705 for availability.
$530.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Continuous-Duty Pumps
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Positive Displacement Pumps
>
Diaphragm Pumps
>
All-Plastic Double-Diaphragm Pumps
> Continuous-Duty Pumps
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title