Cork Borer Set
Cork Borer Set
This chrome-plated steel cork borer set stays sharp. Flat blades permit easy boring of lightweight plastic, cork stoppers, and spongy articles.
Set consists of twelve borers with outside diameters of 4, 6, 7.5, 8.5, 10, 12, 13, 15, 17, 18.5, 19, and 20.5 mm.
 Product Detail
Sort
 Availability
 Price
EW-06298-90 Cork borer set
Cork borer set
Product Type Stopper Hole Borer Sets and Sharpeners,Material Steel,Qty/pk 1
EW-06298-90
In stock.
$84.50 USD/SET of 1
Qty:
See all Cork Borer Set
1 results (showing 1 - 1 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title