log in or register
Learn more:
[-] Show Less
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Crucibles
Crucibles
Cleaning crucibles is essential to maintaining a contaminant free work environment. Often used in ashing and other chemical applications it can be easy for residue to settle on the bottom of crucibles. Whether you are using porcelain, quartz, high-alumnia, platinum or nickel, your crucible will need to be properly cleaned and maintained to continue to serve you well in your applications. Different metals of course require different levels of maintenance. Cleaning crucibles that are platinum require filing them with fused bicarbonate and then boiling them in diluted hydrochloric acid, to remove residue. Cleaning crucibles that are porcelain however just requires the first step, rinse with hot water and clean dry cloth to sweep away the wetness. Though crucibles used in biochemical or pharmaceutical work benefit from sulfuric-nitrate cleaning solutions. Cleaning crucibles need not be difficult but it should be routine.
 Product Detail
Sort
 Availability
 Price
EW-17803-28 Representative Photo Only.  CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 30 mL, 43 mm top OD, 37 mm H, cs of 36
CoorsTek High-Form Crucible, Porcelain; 30 mL, 43 mm top OD, 37 mm H, cs of 36
Product Type Crucibles, High-Form,Capacity mL 30,Material Porcelain
EW-17803-28
In stock.
$254.00 USD/CASE of 36
Qty:
EW-17803-24 Representative Photo Only.  CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 10 mL, 31 mm top OD, 26 mm H, cs of 72
CoorsTek High-Form Crucible, Porcelain; 10 mL, 31 mm top OD, 26 mm H, cs of 72
Product Type Crucibles, High-Form,Capacity mL 10,Material Porcelain
EW-17803-24
In stock.
$301.00 USD/CASE of 72
Qty:
EW-17804-05 Representative photo only. CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, High Alumina; 50ml
CoorsTek High-Form Crucible, High Alumina; 50ml
Product Type Crucibles, High-Form,Capacity mL 50,Material High Alumina
EW-17804-05
In stock.
$50.50 USD/EACH
Qty:
EW-17803-14 Representative Photo Only.  CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 100 mL, 65 mm top OD, 54 mm H, pk of 6
CoorsTek High-Form Crucible, Porcelain; 100 mL, 65 mm top OD, 54 mm H, pk of 6
Product Type Crucibles, High-Form,Capacity mL 100,Material Porcelain
EW-17803-14
In stock.
$62.00 USD/PACKAGE of 6
Qty:
EW-17803-12 Representative Photo Only.  CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 50 mL, 53 mm top OD, 43 mm H, pk of 12
CoorsTek High-Form Crucible, Porcelain; 50 mL, 53 mm top OD, 43 mm H, pk of 12
Product Type Crucibles, High-Form,Capacity mL 50,Material Porcelain
EW-17803-12
In stock.
$103.00 USD/PACKAGE of 12
Qty:
EW-17804-09 Representative photo only. CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, High Alumina; 250ml
CoorsTek High-Form Crucible, High Alumina; 250ml
Product Type Crucibles, High-Form,Capacity mL 250,Material High Alumina
EW-17804-09
In stock.
$112.00 USD/EACH
Qty:
EW-17803-32 Representative Photo Only. CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 50 mL, 53 mm top OD, 43 mm H, cs of 24
CoorsTek High-Form Crucible, Porcelain; 50 mL, 53 mm top OD, 43 mm H, cs of 24
Product Type Crucibles, High-Form,Capacity mL 50,Material Porcelain
EW-17803-32
In stock.
$205.00 USD/CASE of 24
Qty:
Representative photo only Cole Parmer High Form Crucible with Cover Porcelain 25ML
Cole-Parmer High-Form Crucible with Cover, Porcelain; 25ML
Product Type Crucibles, High-Form,Capacity mL 25,Material Porcelain
EW-63100-50
In stock.
$13.00 USD/EACH
Qty:
Representative photo only CoorsTek Wide Form Crucible Porcelain 30 mL 50 mm top OD 31 mm H pk of 12
CoorsTek Wide-Form Crucible, Porcelain; 30 mL, 50 mm top OD, 31 mm H, pk of 12
Product Type Crucibles, Wide-Form,Capacity mL 30,Material Porcelain
EW-17804-06
In stock.
$87.00 USD/PACKAGE of 12
Qty:
EW-17804-07 Representative photo only. CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, High Alumina; 100ml
CoorsTek High-Form Crucible, High Alumina; 100ml
Product Type Crucibles, High-Form,Capacity mL 100,Material High Alumina
EW-17804-07
In stock.
$69.00 USD/EACH
Qty:
EW-07281-56 Representative photo only. Cole-Parmer<small><sup>&reg;</sup></small> High-Form Crucible, Nickel; 100mL (1/pk)
Cole-Parmer High-Form Crucible, Nickel; 100mL (1/pk)
Product Type Crucibles, High-Form,Capacity mL 100,Material Nickel
EW-07281-56
Usually ships in 8 days or sooner.
$56.00 USD/EACH
Qty:
EW-34526-52 Quartz Crucible Only
Quartz Crucible Only
Product Type Crucibles, Standard,Capacity mL 50,Material Quartz
EW-34526-52
In stock.
$111.00 USD/CASE
Qty:
EW-17811-10 Representative Photo Only. CoorsTek<small><sup>&reg;</sup></small> Crucible Cover, Porcelain; 66 mm ID, pk of 6
EW-17811-10
In stock.
$58.00 USD/PACKAGE of 6
Qty:
See all
EW-17803-08 Representative Photo Only.  CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 30 mL, 43 mm top OD, 37 mm H, pk of 12
CoorsTek High-Form Crucible, Porcelain; 30 mL, 43 mm top OD, 37 mm H, pk of 12
Product Type Crucibles, High-Form,Capacity mL 30,Material Porcelain
EW-17803-08
In stock.
$87.00 USD/PACKAGE of 12
Qty:
EW-17803-10 Representative Photo Only.  CoorsTek<small><sup>&reg;</sup></small> High-Form Crucible, Porcelain; 40 mL, 50 mm top OD, 40 mm H, pk of 12
CoorsTek High-Form Crucible, Porcelain; 40 mL, 50 mm top OD, 40 mm H, pk of 12
Product Type Crucibles, High-Form,Capacity mL 40,Material Porcelain
EW-17803-10
In stock.
$91.00 USD/PACKAGE of 12
Qty:
93 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title