United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Category:
Cryo-Glove Cryogenic Gloves
(23)
Cryo-Apron Aprons
(3)
Brand:
Cryo-Glove
(23)
Cryo-Apron
(3)
Product Type:
Gloves
(23)
Aprons
(3)
Price Range:
$100 to $150
(9)
$150 to $200
(17)
Availability:
In Stock
(18)
Material:
Nylon exterior, jersey liner
(23)
Nylon
(3)
Color:
Blue
(3)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Cooling Equipment
>
Cryogenics
> Cryogenic Apparel
Cryogenic Apparel
Choose from our comprehensive line of Cryogenic Apparel and other Cryogenics products.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Water-resistant Cryo-Glove gloves, 15"L, Large, 1 pair
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-14
In stock.
$129.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove Cryogenic Gloves
Waterproof Cryo-Glove gloves, 15" length, medium
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-30
In stock.
$156.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Water-resistant Cryo-Glove gloves, 12"L, large, 1 pair
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-04
In stock.
$119.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Cryo-Apron aprons, 24"W x 42"L
Product Type Aprons,Description Apron,Color Blue
EW-09113-38
In stock.
$156.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Apron Aprons
Waterproof Cryo-Glove gloves, 15" length, large
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-32
In stock.
$156.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Water-resistant Cryo-Glove gloves, 12"L, medium, 1 pair
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-02
In stock.
$119.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Cryo-Apron aprons, 24"W x 36"L
Product Type Aprons,Description Apron,Color Blue
EW-09113-36
In stock.
$145.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Apron
Aprons
Waterproof Cryo-Glove gloves, elbow style, large, 19" length
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-37
In stock.
$187.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Water-resistant Cryo-Glove gloves, 15"L, medium, 1 pair
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-12
In stock.
$129.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Waterproof Cryo-Glove gloves, elbow style, medium, 19" length
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-35
In stock.
$187.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Water-resistant Cryo-Glove gloves, 15"L, X-large, 1 pair
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-16
In stock.
$129.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove Cryogenic Gloves
Cryo-Apron aprons, 24"W x 48"L
Product Type Aprons,Description Apron,Color Blue
EW-09113-40
In stock.
$187.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Apron Aprons
Water-resistant Cryo-Glove gloves, 19" length, medium
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-22
In stock.
$156.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove Cryogenic Gloves
Cryo-Industrial gloves, mid-arm style, large, 15" length
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-64
Usually ships in 4 days.
$162.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove
Cryogenic Gloves
Water-resistant Cryo-Glove gloves, 19" length, large
Product Type Gloves,Material Nylon exterior, jersey liner,Color Blue
EW-09113-24
In stock.
$156.00 USD/PAIR
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Cryo-Glove Cryogenic Gloves
26
results (showing
1
-
15
)
1
2
Next >
>>
Per Page
5
10
15
25
50
Home
>
Cooling Equipment
>
Cryogenics
> Cryogenic Apparel
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title