United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Clean Room
>
Airflow Measurement
>
Anemometers
>
Hot-Wire Anemometers
> Data Logging Hot-Wire Anemometer with CFM/CMM and IR Thermometer
Data Logging Hot-Wire Anemometer with CFM/CMM and IR Thermometer
Built-in infrared thermometer measures noncontact surface temperatures
Five available air speed units ft/min, m/sec, mph, km/hr and knots
Measures air flow volumes up to 2.5 million CFM (72,000 CMM)
Hot-wire sensor handles even the slightest (2 ft./min) air movement with ±3% accuracy
Capture up to 20,000 data points
This hot-wire anemometer has a large backlit LCD that simultaneously displays air flow volume or speed plus remote surface temperature. Ideal for measuring both the volume and temperature of low-speed air flows. A telescoping hot-wire probe extends ot 36" (91.4 cm) for hard-to-reach areas.
The built-in infrared (IR) thermometer allows safe, noncontact measurement of surface temperatures. It offers a distance-to-target ration of 8:1 and emissivity of 0.95 (fixed). Unit allows you to log up to 20,000 data points for download and analysis via USB interface.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Data Logging Hot-Wire Anemometer with CFM/CMM and 8:1 IR Thermometer
Product Type Hot-Wire Anemometer,Display Large, dual backlit LCD,Memory capacity Internal: 9 readings, Min/Max/Avg, data hold
EW-30009-51
Usually ships in 4 days or sooner.
$499.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Data Logging Hot Wire Anemometer with CFM/CMM and IR Thermometer
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Clean Room
>
Airflow Measurement
>
Anemometers
>
Hot-Wire Anemometers
> Data Logging Hot-Wire Anemometer with CFM/CMM and IR Thermometer
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title