United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Moisture Products
>
Moisture Meters
>
Wood / Paper Moisture Meters
> Delmhorst P-2000 Moisture Meter for Paper
Delmhorst P-2000 Moisture Meter for Paper
Ideal for testing paper materials such as paperboard, corrugated stock and paper tubes
Meter measures through built-in pins and optional pin electrodes
This electrical resistance type moisture meter offers three separate scales: paper, baled scrap paper and reference. The moisture scale range for paper is 4.3 to 18%, for baled paper the range is 5 to 40%, and for the reference scale it is 0 to 100. The contact pins mounted on top of the meter provide 5/16'' (0.8 cm) penetration for testing paper tubes or corrugated stock. The meter also features an audible out-of-range alarm, internal calibration check, 100 data point memory, and average/maximum readings.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Delmhorst P-2000 paper moisture meter
Product Type Wood and Paper Moisture Meters,Alarm Yes,Display Digital
EW-59820-41
In stock.
$418.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Delmhorst P-2000 Moisture Meter for Paper
1
results (showing
1
-
1
)
Per Page
5
10
15
25
50
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Moisture Products
>
Moisture Meters
>
Wood / Paper Moisture Meters
> Delmhorst P-2000 Moisture Meter for Paper
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title