Home  >  Lab Equipment > Desiccators
Desiccators
Desiccators
Designed to absorb moisture a desiccant is used in specially-designed, sealable canisters or cabinets that store materials with low-humidity storage requirements. Once the moisture-sensitive object is placed inside the canister with the desiccant it will remain dry. These products are sometimes referred to as desiccant dryers or desiccant dehumidifiers. They come in both non-vacuum and vacuum types. Vacuum desiccators usually have a built-in stopcock which allows the internal air to be evacuated to achieve lower humidity levels. Usually a separate vacuum pump is needed; however the Cole-Parmer®Desi-Vac™ containers feature a convenient built-in pump. These machines are also available with many different sizes, style, color, and feature options. Some even have their own built-in thermohygrometers, ports for flushing the atmosphere with inert gas, and many more advanced features. The desiccant is certainly an important addition to your laboratory.
 Product Detail
Sort
 Availability
 Price
Representative photo only Desiccant Filled Plates 65 mm diameter 6 pack
Desiccant-Filled Plates; 65-mm diameter; 6/pack
Product Type Desiccants,Exterior Dimensions " H 0.55,Exterior Dimensions " D 3.9
EW-07193-95
In stock.
$59.00 USD/PACKAGE of 6
Qty:
EW-07193-05 Indicating Drierite<small><sup>&reg;</sup></small> desiccant; twelve 1-lb packs/carton
Indicating Drierite desiccant; twelve 1-lb packs/carton
Product Type Desiccants,Brand W A HAMMOND DRIERITE CO,Model 23001
EW-07193-05
In stock.
$206.00 USD/CARTON
Qty:
EW-07193-00 Drying column
Drierite Drying column
Product Type Desiccants,Material Polycarbonate,Exterior Dimensions " H 11.375
EW-07193-00
In stock.
$77.00 USD/EACH
Qty:
EW-06520-05 Thermo Scientific Nalgene Vacuum Desiccator, PC cover and body, 280 mm OD
Thermo Scientific Nalgene Vacuum Desiccator, PC cover and body, 280 mm OD
Product Type Vacuum Desiccator,Material polycarbonate,Exterior Dimensions " W 11
EW-06520-05
In stock.
$158.00 USD/EACH
Qty:
EW-08909-52 Dry-Keeper<small><sup>&#153;</sup></small> Desiccator Cabinets, Vertical; Non-Electric
Dry-Keeper Desiccator Cabinets, Vertical; Non-Electric
Product Type Cabinet Desiccator,Exterior Dimensions " W 12.5,Exterior Dimensions " H 20
EW-08909-52
In stock.
$928.00 USD/EACH
Qty:
Representative photo only Indicating Drierite desiccant 25 lb can
Indicating Drierite desiccant; 25-lb can
Product Type Desiccants,Brand W A HAMMOND DRIERITE CO,Model 23025 8 MESH
EW-07193-18
In stock.
$257.00 USD/EACH
Qty:
EW-06514-30 Plastic Vacuum desiccator, 230 mm plate size
Plastic Vacuum desiccator, 230 mm plate size
Product Type Vacuum Desiccator,Material polycarbonate cover, polypropylene body,Exterior Dimensions " W 10.75
EW-06514-30
In stock.
$129.00 USD/EACH
Qty:
Representative photo only Dry Keeper Desiccator Cabinets Vertical Electronic
Dry-Keeper Desiccator Cabinets, Vertical; Electronic
Product Type Cabinet Desiccator,Exterior Dimensions " W 12.5,Exterior Dimensions " H 20
EW-08909-62
In stock.
$1,589.00 USD/EACH
Qty:
Representative photo only Regenerable sponge
Regenerable sponge
Product Type Desiccants,Exterior Dimensions " W 3,Exterior Dimensions " H 0.25
EW-07193-13
In stock.
$42.00 USD/PACKAGE
Qty:
Representative photo only Acrylic Desiccators with Sliding Trays and Built in Hygrometer 9 W x 10 H x 16 D
Acrylic Desiccators with Sliding Trays and Built-in Hygrometer, 9"W x 10-½"H x 16"D
Product Type cabinet Desiccator,Material Acrylic,Exterior Dimensions " W 15.625
EW-08933-30
In stock.
$632.00 USD/EACH
Qty:
EW-79751-30 Vacuum grease
Vacuum Grease, 5.3 Oz
Product Type Vacuum Grease,Bag Size g 150,Brand CONQUEST INC
EW-79751-30
In stock.
$35.00 USD/EACH
Qty:
See all Vacuum Grease
Representative photo only Indicating Drierite desiccant four 5 lb packs carton
Indicating Drierite desiccant; four 5-lb packs/carton
Product Type Desiccants,Brand W A HAMMOND DRIERITE CO,Model 23005
EW-07193-15
In stock.
$250.00 USD/CARTON
Qty:
EW-08900-64 Acrylic Desiccator with gas port
EW-08900-64
Call 1-888-358-4705 for availability.
$1,235.00 USD/EACH
Qty:
EW-08901-35 Electronic desiccator with stainless steel shelves
EW-08901-35
Usually ships in 13 days.
$2,100.00 USD/EACH
Qty:
EW-06536-60 KIMAX<small><sup>&reg;</sup></small> Desiccator, 230 mm
KIMAX Desiccator, 230 mm
Product Type Glass Desiccator,Exterior Dimensions " H 13,Exterior Dimensions " D 9.8
EW-06536-60
In stock.
$479.00 USD/EACH
Qty:
117 results (showing 1 - 15 )
Per Page 
Home  >  Lab Equipment > Desiccators
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title