United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Thermocouple Instruments and Probes
>
Benchtop and Wall Mount Thermocouple Meters
> Digi-Sense Thermocouple Scanning Thermometer with 12-Channels - Benchtop
Digi-Sense Thermocouple Scanning Thermometer with 12-Channels - Benchtop
Store and recall up to 4680 sets of readings with time and date
Automatically or manually display, record and output temperature readings
Field calibrate each probe separately—achieve accuracies of ±0.1% of reading!
Bidirectional RS-232 for setup or PC datalogging
Graph in real timeuse new CD-ROM software (included)
Includes a real-time clock, nonvolatile memory, and parallel printer output
Scan and log up to 12 thermocouple inputs from any combination of J, K, T, E, N, B, R, and S thermocouple probes.
Thermocouple type is individually selectable per channel.
Datalogging feature automatically stores up to 4680 sets of readings.
Download readings or stored data to your computer. Link up to 8 units together to expand channel capacity.
Sequentially captures temperature data, allowing you to monitor and record process variations-ideal for statistical process control.
Easy-to-use front panel MENU key lets you program datalogging and print intervals, scan rate, date and time, temperature scale (degF, degC, K, or degR), resolution (0.1 or 1deg), channels displayed, thermocouple type, and data output parameters.
Each channel features HOLD, STORE, MAX and MIN (measur
When simultaneous monitoring of temperatures during different process steps or phases is critical, these new 12-channel scanning thermometers are ideal. Wall mount unit is ideal for permanent installation and continuous monitoring of your processes. Benchtop unit accepts most any power source giving you great flexibility as to where and when it may be used. Both models function as a clatalogger-store data in the non-volatile memory and later download to your printer or PC. Link both models in series to expand the number of channels you can scan. Each of the 12 thermocouple inputs are programmable to scan from every four seconds to hourly.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Digi-Sense 12 Channel Scanning Benchtop Thermometer, 230 VAC
Product Type Thermocouple meter,Min temperature º F
418,Min temperature º C 3272
EW-92000-05
In stock.
$1,807.95 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Digi-Sense Thermocouple Scanning Thermometer with 12-Channels - Benchtop
Digi-Sense 12 Channel Scanning Benchtop Thermometer, 115 VAC
Product Type Thermocouple meter,Min temperature º F
418,Min temperature º C 3272
EW-92000-00
In stock.
$1,807.95 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Digi-Sense Thermocouple Scanning Thermometer with 12-Channels - Benchtop
2
results (showing
1
-
2
)
Per Page
5
10
15
25
50
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Thermocouple Instruments and Probes
>
Benchtop and Wall Mount Thermocouple Meters
> Digi-Sense Thermocouple Scanning Thermometer with 12-Channels - Benchtop
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title