United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$500 to $799
(2)
$800 to $1,099
(1)
Brix scale range (Maximum):
95
(2)
60
(1)
Refractive Index (Maximum):
1.53
(2)
Refractive Index (Minimum):
1.33
(1)
1.4098
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Lab Equipment
>
Refractometers
>
Digital Benchtop Refractometers
> Digital Laboratory Refractometers
Digital Laboratory Refractometers
Provide process and quality control with ease
Four models cover all major lab refractometer applications
Compact design saves valuable benchtop space
These compact benchtop refractometers are ideal for measuring Brix, salinity, and refractive index with the full scale accuracy of 0.1%. The internal CCD scanner (linear scanned array imaging) provides accurate, reliable measurements. Units are simple to use – place at least 0.4 mL of sample on the surface of the prism, press READ, and the sample results are displayed in seconds. The meter instantly displays the measurement and temperature on the large, easy-to-read LCD.
The sophisticated microprocessor provides automatic calibration, automatic temperature compensation, low-battery indicator, automatic shutoff (after three minutes) and visual hi/low alarms. The durable, water-resistant design protects the electronics in wet environments and makes cleanup easy. All models calibrate to distilled water and feature a simple push-button interface and 5000 reading battery life.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Digital Refractometer, Brix, 0.0 to 95.0%
Product Type Refractometer (Digital),Brix scale range Minimum 0,Brix scale range Maximum 95
EW-02941-34
In stock.
$805.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Digital Laboratory Refractometers
Digital Refractometer, Brix, 0.0 to 60.0%, 0.0 to 28.0% Salinity, 1.330 to 1.4419 RI
Product Type Refractometer (Digital),Brix scale range Minimum 0,Brix scale range Maximum 60
EW-02941-35
In stock.
$585.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Digital Laboratory Refractometers
Digital Refractometer, Brix, 45.0 to 95.0%
Product Type Refractometer (Digital),Brix scale range Minimum 0,Brix scale range Maximum 95
EW-02941-33
In stock.
$630.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Digital Laboratory Refractometers
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Lab Equipment
>
Refractometers
>
Digital Benchtop Refractometers
> Digital Laboratory Refractometers
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title