Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Availability:
Capacity (mL):
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Fluid & Liquid Handling > Burettes (Burets) > Electronic Burettes > Digitrate Electronic Burettes

Digitrate Electronic Burettes

Digitrate Electronic Burettes

  • Glass barrel lets you visually inspect reagent
  • Simple user calibration and factory reset
Digital readout displays meniscus-free total volume readings—using these electronic burettes is so easy. To move from one titration to the next, simply note your final reading, then press the zero switch to continue to the next titration. Low gearing enables dispensing in single increments of 0.01 mL with accuracy better than ±0.2%. The readout is not affected during refilling—dispense up to 99.99 mL without interruption.
Comes with a lifetime battery that powers more than 60,000 titrations. Optional biological filter is sold separately. Order 1400-mL or 2500-mL bottles.
What’s included: dispense nozzle protection sleeve; lithium battery; threaded adapters for 33-, 38-, and 45-mm bottle necks; calibration test certificate.
 Product Detail
Sort
 Availability
 Price
EW-25201-05 Digitrate<small><sup>&reg;</sup></small> Electronic Burettes; 30 mL capacity

Digitrate Electronic Burettes; 30 mL capacity

Product Type Electronic Burettes,Capacity mL 30,Brand Digitrate
EW-25201-05
Usually ships in 8 days.
$952.00 USD/EACH

Qty:

Representative photo only Digitrate Electronic Burettes 50 mL capacity

Digitrate Electronic Burettes; 50 mL capacity

Product Type Electronic Burettes,Capacity mL 50,Brand Digitrate
EW-25201-15
In stock.
$952.00 USD/EACH

Qty:

2 results (showing 1 - 2 )
Per Page 
Home  >  Fluid & Liquid Handling > Burettes (Burets) > Electronic Burettes > Digitrate Electronic Burettes

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title