Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Category:
Brand:
Product Type:
Availability:
Capacity (Liters):
0 to 1 (4)
1 to 5 (1)
Capacity (mL):
0 - 20 (4)
0.2 (2)
Volume (mL):
10 (17)
5 (12)
25 (10)
50 (8)
2 (7)
30 (7)
60 (6)
2.5 (4)
100 (4)
1 (3)
20 (2)
40 (1)
15 (1)
0.5 (1)
5.9 (1)
10.9 (1)
59 (1)
109 (1)
Material:
Glass (5)
Learn more:
[-] Show Less
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
 
Home  >  Lab Equipment > Dispensers

Dispensers

Dispensers

In the laboratory or in the field, the bottle dispenser serves to distribute smaller volumes of reagents and other solutions from large storage bottles and jugs. Essentially, the bottle dispenser consists of a seal, a pump (whether motorized or manual), a volume controller, and a spout. To select the appropriate bottle dispenser, first consider wetted materials; the pump components must be chemically compatible with the bottle’s contents. Many dispensers are mechanized, and all moving parts of the pump and its seals must be taken into account. Volume range, accuracy, and reproducibility all play important roles, too. Commonly, volumes available range from 0.1 to 100 mL. Most offer accuracy of at least +/-1.0% and reproducibility of +/-0.1%. Some bottle dispensers include other features, such as programmable distribution to speed up repeated volume dispensing, or low-profile designs to conserve valuable lab space. For interchangeable use, select a bottle dispenser that is autoclavable.
 Product Detail
Sort
 Availability
 Price
EW-25208-06 Cole-Parmer Bottletop Dispenser 1-10 mL

Cole-Parmer Bottletop Dispenser 1-10 mL

Product Type Dispenser (Bottletop),Volume mL 10,Autoclavable Yes
EW-25208-06
In stock.
$295.00 USD/EACH

Qty:

Representative photo only Cole Parmer Bottletop Dispenser 5 60 mL

Cole-Parmer Bottletop Dispenser 5-60 mL

Product Type Dispenser (Bottletop),Volume mL 60,Autoclavable Yes
EW-25208-10
In stock.
$360.00 USD/EACH

Qty:

Representative photo only Cole Parmer Bottletop Dispenser 2 5 30 mL

Cole-Parmer Bottletop Dispenser 2.5-30 mL

Product Type Dispenser (Bottletop),Volume mL 30,Autoclavable Yes
EW-25208-08
In stock.
$360.00 USD/EACH

Qty:

Representative photo only BrandTech Dispensette Organic Bottletop Dispenser Analog Volume Setting 1 0 to 10 0 mL

BrandTech Dispensette Organic Bottletop Dispenser, Analog Volume Setting, 1.0 to 10.0 mL

Product Type Dispenser (Bottletop),Volume mL 10,Autoclavable Yes
EW-07902-38
In stock.
$465.00 USD/EACH

Qty:

EW-07844-00 07840-00 Gilmont micriometer syringe shown

Gilmont micrometer syringe, 2 mL

Product Type Syringe,Capacity mL 2,Brand Gilmont
EW-07844-00
In stock.
$203.00 USD/EACH

Qty:

Representative photo only Cole Parmer Bottletop Dispenser 2 5 30 mL with 500 mL Glass Bottle

Cole-Parmer Bottletop Dispenser 2.5-30 mL with 500 mL Glass Bottle

Product Type Bottletop Dispensers,Volume mL 30,Autoclavable Yes
EW-25208-18
In stock.
$374.00 USD/EACH

Qty:

EW-07878-06 Adjustable-volume digital dispensers

Adjustable-volume digital dispensers

Product Type Dispenser,Volume mL 10,Autoclavable Yes
EW-07878-06
In stock.
$509.00 USD/EACH

Qty:

Representative photo only Cole Parmer Bottletop Dispenser 5 5 mL

Cole-Parmer Bottletop Dispenser .5-5 mL

Product Type Dispenser (Bottletop),Volume mL 5,Autoclavable Yes
EW-25208-04
In stock.
$295.00 USD/EACH

Qty:

Representative photo only Adjustable Volume analog dispenser 1 10 mL

Adjustable-Volume analog dispenser - 1-10 mL

Product Type Dispenser,Volume mL 10,Autoclavable Yes
EW-07902-18
In stock.
$434.00 USD/EACH

Qty:

Representative photo only BrandTech Seripettor Economy Dispenser 1 0 to 10 mL

BrandTech Seripettor Economy Dispenser, 1.0 to 10 mL

Product Type Dispenser (Bottletop),Volume mL 10,Autoclavable Lower body is autoclavable; pump assembly and cartridge are not
EW-07878-30
In stock.
$138.00 USD/EACH

Qty:

Representative photo only Adjustable Volume analog dispenser 10 100 mL

Adjustable-Volume analog dispenser - 10-100 mL

Product Type Dispenser,Volume mL 100,Autoclavable Yes
EW-07902-26
In stock.
$950.00 USD/EACH

Qty:

Representative photo only BrandTech DispensetteOrganic Bottletop Dispenser Digital Volume Setting 5 to 50 mL

BrandTech DispensetteOrganic Bottletop Dispenser, Digital Volume Setting, 5 to 50 mL

Product Type Dispenser (Bottletop),Volume mL 50,Autoclavable Yes
EW-07878-48
In stock.
$740.00 USD/EACH

Qty:

EW-07912-02 Fixed-Volume Dispenser, 1 mL

Fixed-Volume Dispenser, 1 mL

Product Type Dispenser,Volume mL 1,Autoclavable Yes
EW-07912-02
In stock.
$435.00 USD/EACH

Qty:

Representative photo only Brinkmann ChemSaver Dispenser 1 5 mL

Brinkmann ChemSaver Dispenser 1-5 mL

Product Type Dispenser,Volume mL 5,Autoclavable Yes
EW-07932-01
In stock.
$588.00 USD/EACH

Qty:

Representative photo only Cole Parmer Bottletop Dispenser 25 2 5 mL

Cole-Parmer Bottletop Dispenser .25-2.5 mL

Product Type Dispenser (Bottletop),Volume mL 2.5,Autoclavable Yes
EW-25208-02
In stock.
$295.00 USD/EACH

Qty:

110 results (showing 1 - 15 )
Per Page 
Home  >  Lab Equipment > Dispensers

View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title