Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
 
Home  >  Dry Keeper Desiccator Cabinets

Dry Keeper Desiccator Cabinets

Dry Keeper Desiccator Cabinets
Choose from our comprehensive line of Dry Keeper Desiccator Cabinets and other Desiccator Cabinets products.
 Product Detail
Sort
 Availability
 Price

Dry-Keeper Desiccator Cabinets, Vertical; Non-Electric

  • Product Type: Desiccator
  • Exterior Dimensions: 12.5 " W
  • Exterior Dimensions: 20 " H
EW-08909-52
Usually ships in 8 days or sooner.
$983.00 USD/EACH

Qty:

Dry-Keeper Desiccator Cabinets, Horizontal; Electronic

  • Product Type: Desiccator
  • Exterior Dimensions: 24.5 " W
  • Exterior Dimensions: 13.75 " H
EW-08909-60
Available for order
or call 1-888-358-4705 to order
$1,639.00 USD/EACH

Qty:

Dry-Keeper Desiccator Cabinets, Vertical; Electronic

  • Product Type: Desiccator
  • Exterior Dimensions: 12.5 " W
  • Exterior Dimensions: 20 " H
EW-08909-62
Usually ships in 10 days or sooner.
$1,686.00 USD/EACH

Qty:

Dry-Keeper Plus Desiccator Cabinets, Vertical; Electronic

  • Product Type: Desiccator
  • Exterior Dimensions: 12 " W
  • Exterior Dimensions: 13.875 " H
EW-08909-64
Available for order
or call 1-888-358-4705 to order
$1,874.00 USD/EACH

Qty:

Dry-Keeper Desiccator Cabinets, Horizontal; Non-Electric

  • Product Type: Desiccator
  • Exterior Dimensions: 19.625 " W
  • Exterior Dimensions: 13.75 " H
EW-08909-50
Available for order
or call 1-888-358-4705 to order
$1,333.00 USD/EACH

Qty:

5 results (showing 1 - 5 )
Per Page 
#PRODUCTTITLE#

#PRODUCTHEADER#

#IAMGESTICKER#
#CROSSOUTPRICE#
#PRICE#
#PRICEWITHVAT#
#AVAILABILITY#
Qty
Quantity Discounts
Qty: #QTY10COUNT# #QTY15COUNT# #QTY20COUNT#
Price: #QTY10PRICE# #QTY15PRICE# #QTY20PRICE#
#PRODUCTIMAGE#
#POWERREVIEW#
#MARKETINGTEXT#
#ICONS#
#SPECS#
Home  >  Dry Keeper Desiccator Cabinets