Home  >  Pumps > Metering Pumps > Piston Pumps > Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
  • Ideal for liquid and vapor refrigerant recovery, water sampling/treatment, and volatile chemicals
  • Oil-free and leak-proof
  • Only one moving part and two check valves
  • Compact and quiet (<60 db for liquids)
With the variable speed rate controller, pumps feature multiple control options for adjusting the flow of your fluid or gas. Control options include: manual on/off switch, dry contacts (for remote on/off), 4 to 20 mA (with 79306-75), and PC or PLC interface. Custom control options are available, call our Application Specialists for availability.
Pumps also include integral overload protection, run-dry capability, self-priming up to 22-ft, an integral solid state circuit board (for control of pump cycle rates) and submersible connections.
Alternately energized and de-energized coils located at either end of the pump change the magnetic field surrounding the piston. This results in movement of the piston back and forth within the piston cylinder and hence movement of the fluid. The check valves located in the piston and pump outlet prevent any reverse flow from occurring.
Required system components include: a variable-speed liquid or gas piston pump, a variable-speed rate controller (with or without a 4 to 20 mA input), a signal power cord, and a power cord.
Note: Call our Application Specialists for longer signal and power cord lengths.
 Product Detail
Sort
 Availability
 Price
Representative photo only Variable speed liquid piston pump standard 115 VAC
Variable-speed liquid piston pump, standard, 115 VAC
Product Type Piston Metering Pumps,Max pressure 45 psi (1400 psi for system),Max flow rate mL/min 2,902
EW-79306-00
Usually ships in 14 days.
$1,332.00 USD/EACH
Qty:
Representative photo only Variable speed rate controller standard with 4 to 20 mA input 115 VAC
Variable-speed rate controller, standard with 4 to 20 mA input, 115 VAC
Product Type Piston Metering Pumps,Power VAC 115 (9 VDC adapter is included),Model VRC2001
EW-79306-75
Usually ships in 14 days.
$582.00 USD/EACH
Qty:
Representative photo only Stainless steel strainer 60 mesh 5 8 diameter
Stainless steel strainer; 60 mesh, 5/8" diameter
Product Type Metering Pump Accessories,Model STR 1-4-12
EW-79306-82
Usually ships in 14 days.
$160.00 USD/EACH
Qty:
Representative photo only Stainless steel stand pipe assembly for 30 or 50 gallon drums
Stainless steel stand pipe assembly; for 30 or 50 gallon drums
Product Type Metering Pump Accessories,Model 20065
EW-79306-83
Usually ships in 14 days.
$550.00 USD/EACH
Qty:
Representative photo only Variable speed gas piston pump Class 1 Division 2 rated for hazardous duty locations 115 VAC
Variable-speed gas piston pump, Class 1, Division 2 rated for hazardous duty locations, 115 VAC
Product Type Piston Metering Pumps,Max pressure 130 psi (1400 psi for system),Max flow rate mL/min 66
EW-79306-15
Usually ships in 14 days.
$2,000.00 USD/EACH
Qty:
EW-79306-20 Variable-speed liquid piston pump, Class 1, Division 1 rated for hazardous duty locations, 115 VAC
Variable-speed liquid piston pump, Class 1, Division 1 rated for hazardous duty locations, 115 VAC
Product Type Piston Metering Pumps,Max pressure 45 psi (1400 psi for system),Max flow rate mL/min 2,902
EW-79306-20
Usually ships in 14 days.
$2,250.00 USD/EACH
Qty:
Representative photo only Mounting kit stainless steel
Mounting kit; stainless steel
Product Type Metering Pump Accessories,Model 20051
EW-79306-80
Usually ships in 14 days.
$14.00 USD/EACH
Qty:
Representative photo only Variable speed gas piston pump standard 115 VAC
Variable-speed gas piston pump, standard, 115 VAC
Product Type Piston Metering Pumps,Max pressure 130 psi (1400 psi for system),Max flow rate mL/min 66
EW-79306-05
Usually ships in 14 days.
$1,332.00 USD/EACH
Qty:
Representative photo only Variable speed gas piston pump Class 1 Division 1 rated for hazardous duty locations 115 VAC
Variable-speed gas piston pump, Class 1, Division 1 rated for hazardous duty locations, 115 VAC
Product Type Piston Metering Pumps,Max pressure 130 psi (1400 psi for system),Max flow rate mL/min 66
EW-79306-25
Usually ships in 14 days.
$2,250.00 USD/EACH
Qty:
Representative photo only Stainless steel strainer 60 mesh 1 diameter
Stainless steel strainer; 60 mesh, 1" diameter
Product Type Metering Pump Accessories,Model STR 1-2-23
EW-79306-81
Usually ships in 14 days.
$160.00 USD/EACH
Qty:
EW-79306-70 Variable-speed rate controller, standard, 115 VAC
Variable-speed rate controller, standard, 115 VAC
Product Type Piston Metering Pumps,Power VAC 115 (9 VDC adapter is included),Model VRC2000
EW-79306-70
Usually ships in 14 days.
$500.00 USD/EACH
Qty:
Representative photo only Variable speed liquid piston pump Class 1 Division 2 rated for hazardous duty locations 115 VAC
Variable-speed liquid piston pump, Class 1, Division 2 rated for hazardous duty locations, 115 VAC
Product Type Piston Metering Pumps,Max pressure 45 psi (1400 psi for system),Max flow rate mL/min 2,902
EW-79306-10
Call 1-888-358-4705 for availability.
$2,000.00 USD/EACH
Qty:
Representative photo only Portable frame with SS pump mounting kit
Portable frame; with SS pump mounting kit
Product Type Metering Pump Accessories,Model 20056
EW-79306-84
Usually ships in 14 days.
$90.00 USD/EACH
Qty:
13 results (showing 1 - 13 )
Per Page 
Home  >  Pumps > Metering Pumps > Piston Pumps > Dual Solenoid Variable-speed Liquid and Gas Piston Pumps
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title