Dye Tracers
Dye Tracers
Water tracing dye is a safe, non-toxic and biodegradable way to determine the direction and rate of water flow in various systems. The water tracing dye dissolves rapidly in water, producing a vivid, easy-to-see color used to trace their diffusion. These are excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more. Tracing dyes are EPA-approved and NSF-certified. For opaque water, liquid yellowed or fuzzy with algae growth use the water tracing dye called fluorescent red dye (rhodamine WT) a non-carcinogenic. Use with an ultraviolet lamp (order separately) to trace fluorescent red dyes where normal visual detection is difficult. This type of dye can be used to trace the source of leaks in mechanical and automotive systems like brake lines, transmissions and radiators. Minimum detectable concentration of all dyes in clear water is 1 ppm. Choose from tablet, liquid, or powder forms of dye tracers. Tablets dissolve quickly but are limited to small volume systems. Liquid can be used in most any volume system, but they are the bulkiest in weight and size. Powdered are best used in large volume systems. Find the right water tracing dye in Cole-Parmer’s product listings.
What is pH?
pH is a measure of a liquid’s acidity or alkalinity as given by the relative quantity of hydrogen ions (H+) it contains. The greater the concentration of H+ ions, the more acidic the solution and the lower the pH. Conversely, the lower the H+ concentration, the more basic (or alkaline) the solution and the higher the pH. pH is formally defined as the negative log of H+ activity. This means that every pH unit expresses a tenfold change in acidic strength.
Why is pH important?
pH measurement is critical to a wide variety of industries that affect our everyday lives. Industries such as water and wastewater treatment, chemical production, agriculture, environmental monitoring, life science research, pharmaceutical production, and electronics manufacturing all rely heavily on accurate pH measurement for their processes. Consequently, pH is now the most widely tested chemical parameter in the world.
How do I measure pH?
Quick and dirty pH measurement can be achieved through litmus paper or pH indicators. These are convenient because of their low cost and ease of use, but they can only provide a rough indication of pH, which is insufficient for most applications. The more accurate method involves the use of a measurement system consisting of a pH meter, an electrode that is sensitive to H+ ions, and calibration buffer solutions. The effect that H+ ions have on the electrode can be measured and converted to a pH value by the meter.
Meter Selection
Cole-Parmer has pH meters that are suitable for almost any purpose. Select your meter based on the requirements of your application. Pocket-sized testers are great for their low cost and small size. Handheld, portable meters typically offer more features than testers and are good for laboratory and field use. Benchtop meters are ideal for stationary lab applications typically offering larger displays and more robust data management than other types of meters. ISFET (ionspecific field effect transistor) meters are a unique subset of pH meter that utilize a silicon chip sensor in lieu of a traditional pH-sensitive glass bulb electrodes. This makes them perfect for applications where the presence of glass presents an issue such as with food production.
Electrodes
Most pH measurement systems feature standard pH electrodes with glass bulbs made from pH-sensitive glass, yet there is a wide variety of configurations to handle specialized applications.
Buffers
All pH meters require calibration standards to ensure that the readings are traceable to a standard. Always choose a pH 7 buffer plus at least one other pH value close to your expected measurement range. Using solutions of known pH value allow you to adjust the system to read accurate measurements. Calibration should be performed before each reading or each set of readings.
 Product Detail
Sort
 Availability
 Price
Representative photo only Flourescent FWT Red Dye Concentrate Gallon
Flourescent FWT Red Dye Concentrate, Gallon
Product Type Dye Tracers,Applications Metered systems
EW-00298-06
In stock.
$123.00 USD/GALLON
Qty:
Representative photo only Flourescent FLT Yellow Green Dye Concentrate Gallon
Flourescent FLT Yellow/Green Dye Concentrate, Gallon
Product Type Dye Tracers,Applications Metered systems
EW-00298-07
In stock.
$108.00 USD/GALLON
Qty:
Representative photo only Flourescent FLT Yellow Green Dye Tablets 200 tablets
Flourescent FLT Yellow/Green Dye Tablets, 200 tablets
Product Type Dye Tracers,Applications Plumbing, storm and sanitary drain analysis
EW-00295-17
In stock.
$45.00 USD/EACH
Qty:
Representative photo only Flourescent FWT Red Dye Tablets 200 tablets
Flourescent FWT Red Dye Tablets, 200 tablets
Product Type Dye Tracers,Applications Plumbing, storm and sanitary drain analysis
EW-00295-16
In stock.
$48.50 USD/EACH
Qty:
Representative photo only Flourescent FWT Red Dye Powder 1 Lb
Flourescent FWT Red Dye Powder, 1 Lb
Product Type Dye Tracers,Applications Cooling towers, lakes, rivers, ponds, power plants
EW-00298-26
In stock.
$144.00 USD/EACH
Qty:
Representative photo only Blue Dye Concentrate Gallon
Blue Dye Concentrate, Gallon
Product Type Dye Tracers,Applications Metered systems
EW-00298-08
In stock.
$108.00 USD/GALLON
Qty:
Representative photo only Flourescent FLT Yellow Green Dye Powder 1 Lb
Flourescent FLT Yellow/Green Dye Powder, 1 Lb
Product Type Dye Tracers,Applications Cooling towers, lakes, rivers, ponds, power plants
EW-00298-27
In stock.
$111.00 USD/EACH
Qty:
Representative photo only Flourescent FWT Red Dye Concentrate Pint
Flourescent FWT Red Dye Concentrate, Pint
Product Type Dye Tracers,Applications Squirt-top dispensing
EW-00298-16
In stock.
$27.00 USD/PINT
Qty:
Representative photo only Blue Dye Tablets 200 tablets
Blue Dye Tablets, 200 tablets
Product Type Dye Tracers,Applications Brighten murky ponds
EW-00295-18
In stock.
$55.50 USD/EACH
Qty:
Representative photo only Blue Dye Concentrate Pint
Blue Dye Concentrate, Pint
Product Type Dye Tracers,Applications Squirt-top dispensing
EW-00298-18
In stock.
$26.50 USD/PINT
Qty:
Representative photo only Flourescent FLT Orange Dye Powder 1 Lb
Flourescent FLT Orange Dye Powder, 1 Lb
Product Type Dye Tracers,Applications Cooling towers, lakes, rivers, ponds, power plants
EW-00298-29
In stock.
$144.00 USD/EACH
Qty:
Representative photo only Flourescent FLT Yellow Green Dye Concentrate Pint
Flourescent FLT Yellow/Green Dye Concentrate, Pint
Product Type Dye Tracers,Applications Squirt-top dispensing
EW-00298-17
In stock.
$26.50 USD/PINT
Qty:
Representative photo only Flourescent FLT Orange Dye Tablets 200 tablets
Flourescent FLT Orange Dye Tablets, 200 tablets
Product Type Dye Tracers,Applications Water tracing and leak detection studies
EW-00295-19
In stock.
$46.00 USD/EACH
Qty:
Representative photo only Flourescent FLT Orange Dye Concentrate Pint
Flourescent FLT Orange Dye Concentrate, Pint
Product Type Dye Tracers,Applications Metered systems
EW-00298-19
In stock.
$26.50 USD/PINT
Qty:
EW-00298-28 Blue Dye Powder, 1 Lb
Blue Dye Powder, 1 Lb
Product Type Dye Tracers,Applications Cooling towers, lakes, streams, power plants
EW-00298-28
Usually ships in 7 days.
$111.00 USD/EACH
Qty:
24 results (showing 1 - 15 )
Per Page 
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title