United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$300 to $350
(1)
$400 to $450
(2)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Clean Room
>
Cleanroom Furniture
> ESD Polyurethane Cleanroom Chairs
ESD Polyurethane Cleanroom Chairs
Ideal for use in semiconductor, bioscience, pharmaceutical, and board manufacturing
All chairs feature heavy-duty construction, easy-to-use pneumatic height adjustment, and 360° swivel. Each chair has a sturdy five-legged base with glides; order optional caster separately below. Cleanroom compliance is certified by an independent testing laboratory.
ESD Polyurethane Chairs
have a unique ergonomic design to reduce worker fatigue and improve productivity. Rugged, curved back and tilting, articulating seat provide excellent support and maximum comfort. Also includes conductive polyurethane upholstery, a brass drag chain, and ESD glides.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Cleanroom Chair, ESD Polyurethane, 22" to 32" height, aluminum base
Product Type ESD Cleanroom Chair,Brand Bevco
EW-86309-35
Call 1-888-358-4705 for availability.
$434.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ESD Polyurethane Cleanroom Chairs
Cleanroom Chair, ESD Polyurethane, 19" to 26.5" height, aluminum base
Product Type ESD Cleanroom Chair,Brand Bevco
EW-86309-34
Call 1-888-358-4705 for availability.
$434.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ESD Polyurethane Cleanroom Chairs
Cleanroom Chair, ESD Polyurethane, 16" to 21" height, aluminum base
Product Type ESD Cleanroom Chair,Brand Bevco
EW-86309-33
Call 1-888-358-4705 for availability.
$336.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
ESD Polyurethane Cleanroom Chairs
3
results (showing
1
-
3
)
Per Page
5
10
15
25
50
Home
>
Clean Room
>
Cleanroom Furniture
> ESD Polyurethane Cleanroom Chairs
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title