log in or register
Price Range:
Availability:
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Home  >  Safety Products > Storage and Handling > Safety Cabinets > Eagle PE Acid and Corrosive Cabinets
Eagle PE Acid and Corrosive Cabinets
Eagle PE Acid and Corrosive Cabinets
  • Suitable for storing concentrated sulphuric, hydrocloric, and nitric acid
Constructed of 100% polyethylene, these nonmetallic cabinets are ideal for storing your aggressive chemicals. Adjustable shelves contain small spills and remove for easy cleaning. Doors feature integral handles with key lock holes (lock not included) and recessed areas for additional storage of smaller bottles. Cabinets have a one-gallon top and internal sump capacity, two 2" (5.1 cm) vents with cap, leveling feet, and “Caution Corrosives” label.
 Product Detail
Sort
 Availability
 Price
EW-81773-00 Acid And Corrosive Cabinets, 4 Gal, Blue
Eagle PE Acid And Corrosive Cabinets, 4 Gal, Blue
Product Type Acid Storage Cabinets,Description Benchtop,Capacity gal. 4,Color Blue
EW-81773-00
In stock.
$515.00 USD/EACH
Qty:
EW-81773-04 Acid And Corrosive Cabinets, 44 Gal, Blue
Eagle PE Acid And Corrosive Cabinets, 44 Gal, Blue
Product Type Acid Storage Cabinets,Description Full-size,Capacity gal. 44,Color Blue
EW-81773-04
Usually ships in 13 days.
$1,311.00 USD/EACH
Qty:
Representative photo only Eagle PE Acid And Corrosive Cabinets 22 Gal Blue
Eagle PE Acid And Corrosive Cabinets, 22 Gal, Blue
Product Type Acid Storage Cabinets,Description Under-counter,Capacity gal. 22,Color Blue
EW-81773-02
Usually ships in 10 days.
$1,006.00 USD/EACH
Qty:
Representative photo only Eagle PE Acid And Corrosive Cabinets 4 Gal White
Eagle PE Acid And Corrosive Cabinets, 4 Gal, White
Product Type Acid Storage Cabinets,Description Benchtop,Capacity gal. 4,Color White
EW-81773-06
Call 1-888-358-4705 for availability.
$515.00 USD/EACH
Qty:
Representative photo only Eagle PE Acid And Corrosive Cabinets 44 Gal White
Eagle PE Acid And Corrosive Cabinets, 44 Gal, White
Product Type Acid Storage Cabinets,Description Full-size,Capacity gal. 44,Color White
EW-81773-10
Usually ships in 6 days.
$1,311.00 USD/EACH
Qty:
Representative photo only Eagle PE Acid And Corrosive Cabinets 22 Gal White
Eagle PE Acid And Corrosive Cabinets, 22 Gal, White
Product Type Acid Storage Cabinets,Description Under-counter,Capacity gal. 22,Color White
EW-81773-08
Call 1-888-358-4705 for availability.
$1,006.00 USD/EACH
Qty:
6 results (showing 1 - 6 )
Per Page 
Home  >  Safety Products > Storage and Handling > Safety Cabinets > Eagle PE Acid and Corrosive Cabinets
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title