United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
Price Range:
$500 to $799
(2)
$1,100 to $1,999
(4)
$2,000 and Over
(3)
CE Compliance:
Yes
(7)
Max system pressure (psi):
230
(7)
125
(2)
Max system pressure (bar):
15.8
(5)
8.6
(4)
Motor (VAC):
208-230/460
(5)
115/230
(2)
230/460
(2)
Max flow rate (GPM):
80
(2)
240
(2)
115
(1)
[+]
Show more
(4)
140
(1)
160
(1)
380
(1)
370
(1)
[-]
Show less
Max head (ft):
75
(2)
103
(1)
139
(1)
[+]
Show more
(5)
205
(1)
280
(1)
195
(1)
128
(1)
125
(1)
[-]
Show less
Max flow rate (LPM):
1400
(1)
757
(1)
Max head (m):
38.1
(1)
39
(1)
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Home
>
Pumps
>
Centrifugal Pumps
>
Centrifugal Pumps that are Mechanically Coupled (have a Mechanical Seal)
> Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Rugged continuous-duty service with chemical and corrosion resistance
Close-coupled design saves space; simplifies maintenance and installation
Stainless steel liquid end components deliver corrosion resistance
Versatile mounting can be installed horizontally or vertically
Back-pullout construction assembly allows overhaul of the impeller and seal without disturbing suction
and discharge connections
Top centerline discharge and foot support under casing ensures self-venting and reduces misalignment
from pipe loads
High operating efficiency lowers operating costs
Applications:
• Plant services
• Water supply systems
• Washing plants
• Cooling water
• Car wash
• Scrubbers
• Ultrapure water systems
• Jockey pump services
• Air conditioning
• Sprinkler/flow irrigation
• OEM equipment application
• Pressure boosting
• Liquid transfer
• Heat exchanger
• Spray systems, heating
• Beverage processing
• Pharmaceutical services
• Water reclamation and treatment
• General pump applications
Flange kits include:
stainless steel 150# threaded RF companion flanges, gaskets, nuts, bolts, flat washer, and lockwashers.
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
Compare
Product Detail
Sort
Most Popular
Name
Name Descending
Price (Lowest First)
Price (Highest First)
Availability
Price
Mechanically Coupled 304 SS Pump, High-Head, 160 GPM, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 160,Max head ft 205
EW-70725-31
Usually ships in 7 days.
$2,034.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Mechanically Coupled 304 SS Pump, High-Head, 115 GPM, 115/230 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 115,Max head ft 103
EW-70725-21
Usually ships in 7 days.
$1,900.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Mechanically Coupled 304 SS Pump, High-Head, 240 GPM, 15 Hp, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 240,Max head ft 280
EW-70725-41
Usually ships in 7 days.
$2,574.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Mechanically Coupled 304 SS Pump, High-Head, 380 GPM, 7-1/2 Hp, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 370,Max flow rate LPM 1400
EW-70725-46
Usually ships in 7 days.
$1,962.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Mechanically Coupled 304 SS Pump, High-Head, 240 GPM, 5 Hp, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 240,Max flow rate LPM 757
EW-70725-36
Usually ships in 7 days.
$1,593.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
304 Stainless Steel Pump, 80 GPM, 115/230 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 80,Max head ft 75
EW-70725-11
Usually ships in 7 days.
$773.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Mechanically Coupled 304 SS Pump, High-Head, 380 GPM, 15 Hp, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 380,Max head ft 195
EW-70725-51
Usually ships in 7 days.
$2,671.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
Mechanically Coupled 304 SS Pump, High-Head, 140 GPM, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 140,Max head ft 139
EW-70725-26
Usually ships in 8 days.
$1,655.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
304 Stainless Steel Pump, 80 GPM, 208-230/460 VAC
Product Type Mechanically Coupled Centrifugal Pumps,Max flow rate GPM 80,Max head ft 75
EW-70725-13
Usually ships in 7 days.
$748.00 USD/EACH
Qty:
Add to Cart
Save to shopping list
Accessories
Calibrations
More Details
See all
Ebara 304 SS Mechanically-Coupled Pumps, High-Head
9
results (showing
1
-
9
)
Per Page
5
10
15
25
50
Home
>
Pumps
>
Centrifugal Pumps
>
Centrifugal Pumps that are Mechanically Coupled (have a Mechanical Seal)
> Ebara 304 SS Mechanically-Coupled Pumps, High-Head
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title